Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083067.4 | XP_001083067.1 | 725 | 87 | ||
GenBank mRNA | JV627893.1 | - | 1 - 2 | 312 | 87 | |
JV695486.1 | - | 1 - 2 | 196 | 87 |
Set Membership: |
![]() ![]() ![]() ![]() |