Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194264.2 | NP_001181193.1 | 2 - 3 | 207 | 65 | |
GenBank mRNA | JU472337.1 | - | 2 - 3 | 154 | 65 | |
JV045121.1 | - | 2 - 3 | 202 | 65 | ||
JV453052.1 | - | 2 - 3 | 215 | 65 | ||
JV635645.1 | - | 2 - 3 | 192 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() |