Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198722.1 | NP_001185651.1 | 2 - 3 | 375 | 73 | |
GenBank mRNA | CN641586.1 | - | 2 - 3 | 375 | 73 | |
JU322453.1 | - | 2 - 3 | 413 | 73 | ||
JU472473.1 | - | 2 - 3 | 400 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |