Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC106999105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141765.2 | XP_014997251.1 | 865 | 87 |
Set Membership: |
![]() ![]() ![]() ![]() |