Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00437040_m1
          See other THPO GE Assays ›
          Gene Symbol:
          Thpo LOC103690065
          Gene Name:
          thrombopoietin
          thrombopoietin
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.11: 83868603 - 83873910 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 82846293 - 82852513 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00437040_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thpo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031133.2 NP_112395.1 397 72
          XM_008768827.1 XP_008767049.1 4 - 5 1359 72
          XM_008768828.1 XP_008767050.1 4 - 5 717 72
          XM_008768829.1 XP_008767051.1 3 - 4 1707 72
          GenBank mRNA D32207.1 - 398 72
          Gene Symbol: LOC103690065
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006248552.2 XP_006248614.1 4 - 5 1528 72
          XM_006248553.2 XP_006248615.1 4 - 5 1528 72
          XM_008768815.1 XP_008767037.1 4 - 5 1359 72
          XM_008768816.1 XP_008767038.1 4 - 5 717 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000906.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thpo
          Click here to find siRNAs for Gene LOC103690065

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          cell population proliferation
          positive regulation of cell population proliferation
          myeloid cell differentiation
          response to lipopolysaccharide
          thrombopoietin-mediated signaling pathway
          positive regulation of megakaryocyte differentiation
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          signaling receptor binding
          cytokine activity
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P008879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388327 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008879 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008879 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P008879 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P708819 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701639471 - -
          Agilent Rat cDNA Microarray (G4105A) 701639471 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10752105 - -
          Affymetrix Rat Gene 1.0 ST Array 10752102 - -
          Affymetrix Rat Gene 1.0 ST Array 10752103 - -
          Affymetrix Rat Gene 1.0 ST Array 10752108 - -
          Affymetrix Rat Gene 1.0 ST Array 10752107 - -
          Affymetrix Rat Gene 1.0 ST Array 10752106 - -
          Affymetrix Rat Gene 1.1 ST Array 10752100 - -
          Rat Genome 230 2.0 Array 1387598_at - -
          Rat Genome 230 2.0 Array 1387598_PM_at - -
          Affymetrix HT Rat Focus Array 1387598_PM_at - -
          Affymetrix HT Rat Focus Array 1387598_at - -
          Affymetrix Rat Clariom S Assay PSR1100005104.rn.1 TC1100000906.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100005105.rn.1 TC1100000906.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100005107.rn.1 TC1100000906.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100005111.rn.1 TC1100000906.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100005106.rn.1 TC1100000906.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100005108.rn.1 TC1100000906.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100005110.rn.1 TC1100000906.rn.1 RTA TC
          Rat Expression Set 230 1387598_at - -
          Rat Expression Set 230 1387598_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10752100 - -
          Rat Genome U34 Set D32207_at - -
          Rat Genome U34 Set E12181cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22334352 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366364 - -
          RatRef-12 Expression BeadChip ILMN_55513 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c8579b687-54cxl:80/100.66.135.11:80.
          git-commit: 8e87205ac1279514f9c7a0d34bcd4a43061862b7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0