Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561892_m1
          See other SCNN1B GE Assays ›
          Gene Symbol:
          Scnn1b
          Gene Name:
          sodium channel, non-voltage-gated 1, beta subunit
          Gene Aliases:
          RNENACB
          Chromosome Location:
          Chr.1: 191829552 - 191883991 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561892_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scnn1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012648.1 NP_036780.1 4 - 5 822 84
          XM_008759682.1 XP_008757904.1 4 - 5 857 84
          XM_008759683.1 XP_008757905.1 5 - 6 1067 84
          XM_008759684.1 XP_008757906.1 4 - 5 899 84
          XM_008759685.1 XP_008757907.1 4 - 5 857 84
          XM_008759686.1 XP_008757908.1 4 - 5 857 84
          GenBank mRNA EU627516.1 - 3 - 4 767 84
          U35174.1 - 3 - 4 777 84
          U35175.1 - 3 - 4 777 84
          X77932.1 - 4 - 5 839 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002524.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scnn1b

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of sodium ion transport
          sodium ion transport
          wound healing, spreading of epidermal cells
          sodium ion transmembrane transport
          multicellular organismal water homeostasis
          sensory perception of taste
          sodium ion homeostasis
          sodium channel activity
          protein binding
          ligand-gated sodium channel activity
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170394 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55641 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23752 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068913 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068913 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068913 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P817086 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18475 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50944 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56411 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18331 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710598 - -
          Affymetrix Rat Gene 1.0 ST Array 10710610 - -
          Affymetrix Rat Gene 1.0 ST Array 10710596 - -
          Affymetrix Rat Gene 1.0 ST Array 10710599 - -
          Affymetrix Rat Gene 1.0 ST Array 10710602 - -
          Affymetrix Rat Gene 1.0 ST Array 10710597 - -
          Affymetrix Rat Gene 1.0 ST Array 10710608 - -
          Affymetrix Rat Gene 1.0 ST Array 10710603 - -
          Affymetrix Rat Gene 1.0 ST Array 10710606 - -
          Affymetrix Rat Gene 1.0 ST Array 10710607 - -
          Affymetrix Rat Gene 1.0 ST Array 10710605 - -
          Affymetrix Rat Gene 1.0 ST Array 10710609 - -
          Affymetrix Rat Gene 1.0 ST Array 10710600 - -
          Affymetrix Rat Gene 1.0 ST Array 10710604 - -
          Affymetrix Rat Gene 1.0 ST Array 10710601 - -
          Affymetrix Rat Gene 1.1 ST Array 10710595 - -
          Rat Genome 230 2.0 Array 1370482_PM_at - -
          Rat Genome 230 2.0 Array 1370482_at - -
          Affymetrix HT Rat Focus Array 1370482_at - -
          Affymetrix HT Rat Focus Array 1370482_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100019468.rn.1 TC0100002524.rn.1 RTA TC
          Rat Expression Set 230 1370482_at - -
          Rat Expression Set 230 1370482_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10710595 - -
          Rat Genome U34 Set X77932_at - -
          Rat Genome U34 Set X77932_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21855409 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349849 - -
          RatRef-12 Expression BeadChip ILMN_53253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0