Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567500_m1
          See other INHBA GE Assays ›
          Gene Symbol:
          Inhba
          Gene Name:
          inhibin beta-A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 51894598 - 51920674 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567500_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Inhba
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017128.2 NP_058824.1 1 - 2 60 89
          XM_006254001.2 XP_006254063.1 2 - 3 3719 89
          XM_008771713.1 XP_008769935.1 3 - 4 5427 89
          XM_008771714.1 XP_008769936.1 3 - 4 2207 89
          XM_008771715.1 XP_008769937.1 3 - 4 2109 89
          XM_008771716.1 XP_008769938.1 3 - 4 487 89
          GenBank mRNA M37482.1 - 1 - 2 55 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001906.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the M37482.1, NM_017128.2, XM_006254001.2, XM_008771713.1, XM_008771714.1, XM_008771715.1, XM_008771716.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Inhba

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          ovarian follicle development
          mesoderm formation
          hair follicle development
          hematopoietic progenitor cell differentiation
          regulation of transcription by RNA polymerase II
          negative regulation of cell population proliferation
          male gonad development
          response to xenobiotic stimulus
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of cell growth
          positive regulation of cellular protein metabolic process
          activin receptor signaling pathway
          endodermal cell differentiation
          growth
          odontogenesis
          hemoglobin biosynthetic process
          progesterone secretion
          regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of erythrocyte differentiation
          negative regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of follicle-stimulating hormone secretion
          mesodermal cell differentiation
          cell development
          roof of mouth development
          positive regulation of ovulation
          SMAD protein signal transduction
          eyelid development in camera-type eye
          cellular response to follicle-stimulating hormone stimulus
          cellular response to cholesterol
          extrinsic apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          signaling receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          hormone activity
          growth factor activity
          peptide hormone binding
          inhibin binding
          identical protein binding
          protein heterodimerization activity
          type II activin receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1700012864.rn.1 TC1700001906.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012863.rn.1 TC1700001906.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0