Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569848_m1
          See other TLR4 GE Assays ›
          Gene Symbol:
          Tlr4
          Gene Name:
          toll-like receptor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 82587424 - 82601056 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569848_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tlr4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019178.1 NP_062051.1 2 - 3 492 127
          GenBank mRNA AF057025.1 - 2 - 3 514 127
          EU929065.1 - 2 - 3 217 127
          GQ175880.1 - 2 - 3 492 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000802.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Rat Chemokine Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tlr4

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          response to hypoxia
          microglial cell activation
          activation of innate immune response
          toll-like receptor signaling pathway
          B cell proliferation involved in immune response
          nitric oxide production involved in inflammatory response
          MyD88-dependent toll-like receptor signaling pathway
          innate immune response-activating signal transduction
          skeletal muscle contraction
          leukotriene metabolic process
          inflammatory response
          response to oxidative stress
          I-kappaB phosphorylation
          response to bacterium
          response to toxic substance
          positive regulation of platelet activation
          response to activity
          immunoglobulin mediated immune response
          positive regulation of B cell proliferation
          lipopolysaccharide-mediated signaling pathway
          response to lipopolysaccharide
          detection of lipopolysaccharide
          response to retinoic acid
          response to progesterone
          interferon-gamma production
          negative regulation of interferon-gamma production
          negative regulation of interleukin-17 production
          negative regulation of interleukin-23 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of chemokine production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          response to insulin
          positive regulation of stress-activated MAPK cascade
          toll-like receptor 4 signaling pathway
          macrophage activation
          positive regulation of NF-kappaB import into nucleus
          positive regulation of tumor necrosis factor biosynthetic process
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to morphine
          positive regulation of DNA binding
          positive regulation of interleukin-12 biosynthetic process
          innate immune response
          positive regulation of MHC class II biosynthetic process
          positive regulation of interferon-beta biosynthetic process
          positive regulation of interleukin-8 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          response to ethanol
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of lymphocyte proliferation
          regulation of cytokine secretion
          regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-negative bacterium
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          response to glucocorticoid
          positive regulation of nitric-oxide synthase biosynthetic process
          regulation of sensory perception of pain
          intestinal epithelial structure maintenance
          positive regulation of macrophage cytokine production
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway
          positive regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
          response to fatty acid
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to mechanical stimulus
          cellular response to retinoic acid
          response to gold nanoparticle
          lipopolysaccharide binding
          lipopolysaccharide receptor activity
          receptor activity
          transmembrane signaling receptor activity
          protein binding
          phosphatidylinositol 3-kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P996124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59946 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P996124 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P996124 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P996124 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P996124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60917 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10869538 - -
          Affymetrix Rat Gene 1.0 ST Array 10869536 - -
          Affymetrix Rat Gene 1.0 ST Array 10869537 - -
          Affymetrix Rat Gene 1.1 ST Array 10869535 - -
          Rat Genome 230 2.0 Array 1387982_at - -
          Rat Genome 230 2.0 Array 1387982_PM_at - -
          Affymetrix HT Rat Focus Array 1387982_at - -
          Affymetrix HT Rat Focus Array 1387982_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500005378.rn.1 TC0500000802.rn.1 RTA TC
          Rat Expression Set 230 1387982_PM_at - -
          Rat Expression Set 230 1387982_at - -
          Affymetrix Rat Gene 1.0 ST Array 10869535 - -
          Rat Genome U34 Set rc_AI030997_s_at - -
          Rat Genome U34 Set AF057025_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21139849 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364385 - -
          RatRef-12 Expression BeadChip ILMN_62916 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix