Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581237_m1
          See other FUT2 GE Assays ›
          Gene Symbol:
          Fut2
          Gene Name:
          fucosyltransferase 2 (secretor status included)
          Gene Aliases:
          Ftc, Futb
          Chromosome Location:
          Chr.1: 101631633 - 101652926 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581237_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fut2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031635.1 NP_113823.1 2 - 3 206 71
          XM_006229130.2 XP_006229192.1 1 - 2 102 71
          XM_006229135.2 XP_006229197.1 2 - 3 194 71
          XM_006229136.2 XP_006229198.1 1 - 2 105 71
          XM_008759412.1 XP_008757634.1 1 - 2 213 71
          GenBank mRNA AB006138.1 - 2 - 3 206 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005402.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the AB006138.1, NM_031635.1, XM_006229130.2, XM_006229135.2, XM_006229136.2, XM_008759412.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fut2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          fucosylation
          L-fucose catabolic process
          galactoside 2-alpha-L-fucosyltransferase activity
          fucosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P667763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166974 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105359 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P667763 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P667763 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P667763 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P667763 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P262324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80814 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48428 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721876 - -
          Affymetrix Rat Gene 1.0 ST Array 10721877 - -
          Affymetrix Rat Gene 1.0 ST Array 10721878 - -
          Affymetrix Rat Gene 1.0 ST Array 10721879 - -
          Affymetrix Rat Gene 1.1 ST Array 10721875 - -
          Rat Genome 230 2.0 Array 1381638_PM_at - -
          Rat Genome 230 2.0 Array 1381638_at - -
          Rat Genome 230 2.0 Array 1387097_PM_at - -
          Rat Genome 230 2.0 Array 1371216_PM_s_at - -
          Rat Genome 230 2.0 Array 1369916_at - -
          Rat Genome 230 2.0 Array 1387097_at - -
          Rat Genome 230 2.0 Array 1371215_PM_at - -
          Rat Genome 230 2.0 Array 1371216_s_at - -
          Rat Genome 230 2.0 Array 1371215_at - -
          Rat Genome 230 2.0 Array 1369916_PM_at - -
          Affymetrix HT Rat Focus Array 1371216_s_at - -
          Affymetrix HT Rat Focus Array 1371215_at - -
          Affymetrix HT Rat Focus Array 1371215_PM_at - -
          Affymetrix HT Rat Focus Array 1387097_PM_at - -
          Affymetrix HT Rat Focus Array 1387097_at - -
          Affymetrix HT Rat Focus Array 1371216_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0100044144.rn.1 TC0100005402.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100044142.rn.1 TC0100005402.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100044145.rn.1 TC0100005402.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100044146.rn.1 TC0100005402.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100044148.rn.1 TC0100005402.rn.1 RTA TC
          Rat Expression Set 230 1369916_at - -
          Rat Expression Set 230 1387097_at - -
          Rat Expression Set 230 1369916_PM_at - -
          Rat Expression Set 230 1371216_PM_s_at - -
          Rat Expression Set 230 1371215_PM_at - -
          Rat Expression Set 230 1371215_at - -
          Rat Expression Set 230 1387097_PM_at - -
          Rat Expression Set 230 1371216_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721875 - -
          Rat Genome U34 Set AB006138_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21274618 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358838 - -
          RatRef-12 Expression BeadChip ILMN_60118 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0