Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582138_m1
          See other GGCX GE Assays ›
          Gene Symbol:
          Ggcx
          Gene Name:
          gamma-glutamyl carboxylase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 100277383 - 100293097 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582138_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ggcx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031756.1 NP_113944.1 6 - 7 800 67
          GenBank mRNA AF065387.1 - 6 - 7 800 67
          BC168647.1 - 6 - 7 771 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001122.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ggcx

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-glutamic acid carboxylation
          response to vitamin K
          gamma-glutamyl carboxylase activity
          vitamin binding
          peptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P115483 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P145166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167873 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P145166 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115483 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115483 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P145166 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P145166 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P115483 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P249092 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12664 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700636454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86489 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46731 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77849 - -
          Agilent Rat cDNA Microarray (G4105A) 700636454 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856410 - -
          Affymetrix Rat Gene 1.0 ST Array 10856395 - -
          Affymetrix Rat Gene 1.0 ST Array 10856402 - -
          Affymetrix Rat Gene 1.0 ST Array 10856400 - -
          Affymetrix Rat Gene 1.0 ST Array 10856409 - -
          Affymetrix Rat Gene 1.0 ST Array 10856406 - -
          Affymetrix Rat Gene 1.0 ST Array 10856403 - -
          Affymetrix Rat Gene 1.0 ST Array 10856397 - -
          Affymetrix Rat Gene 1.0 ST Array 10856411 - -
          Affymetrix Rat Gene 1.0 ST Array 10856412 - -
          Affymetrix Rat Gene 1.0 ST Array 10856398 - -
          Affymetrix Rat Gene 1.0 ST Array 10856407 - -
          Affymetrix Rat Gene 1.0 ST Array 10856396 - -
          Affymetrix Rat Gene 1.0 ST Array 10856405 - -
          Affymetrix Rat Gene 1.0 ST Array 10856408 - -
          Affymetrix Rat Gene 1.0 ST Array 10856401 - -
          Affymetrix Rat Gene 1.0 ST Array 10856413 - -
          Affymetrix Rat Gene 1.0 ST Array 10856404 - -
          Affymetrix Rat Gene 1.0 ST Array 10856399 - -
          Affymetrix Rat Gene 1.0 ST Array 10856414 - -
          Affymetrix Rat Gene 1.1 ST Array 10856394 - -
          Rat Genome 230 2.0 Array 1368314_PM_at - -
          Rat Genome 230 2.0 Array 1368314_at - -
          Affymetrix HT Rat Focus Array 1368314_PM_at - -
          Affymetrix HT Rat Focus Array 1368314_at - -
          Affymetrix Rat Clariom S Assay PSR0400008048.rn.1 TC0400001122.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400008049.rn.1 TC0400001122.rn.1 RTA TC
          Rat Expression Set 230 1368314_PM_at - -
          Rat Expression Set 230 1368314_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856394 - -
          Rat Genome U34 Set AF065387_at - -
          Rat Genome U34 Set AF065387_g_at - -
          Rat Genome U34 Set rc_AI137138_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22407543 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364162 - -
          RatRef-12 Expression BeadChip ILMN_67463 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix