Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584554_m1
          See other SMPX GE Assays ›
          Gene Symbol:
          Smpx
          Gene Name:
          small muscle protein, X-linked
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 40030244 - 40086870 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584554_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smpx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053395.1 NP_445847.1 2 - 3 226 64
          XM_006256945.2 XP_006257007.1 2 - 3 195 64
          GenBank mRNA AF364071.1 - 2 - 3 226 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001711.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smpx

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180117 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90367 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14371 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14371 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10789 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31991 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62247 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90087 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938065 - -
          Affymetrix Rat Gene 1.0 ST Array 10938063 - -
          Affymetrix Rat Gene 1.0 ST Array 10938061 - -
          Affymetrix Rat Gene 1.0 ST Array 10938062 - -
          Affymetrix Rat Gene 1.0 ST Array 10938064 - -
          Affymetrix Rat Gene 1.1 ST Array 10938060 - -
          Rat Genome 230 2.0 Array 1370165_at - -
          Rat Genome 230 2.0 Array 1370165_PM_at - -
          Affymetrix HT Rat Focus Array 1370165_at - -
          Affymetrix HT Rat Focus Array 1370165_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00011128.rn.1 TC0X00001711.rn.1 RTA TC
          Rat Expression Set 230 1370165_PM_at - -
          Rat Expression Set 230 1370165_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938060 - -
          Rat Genome U34 Set rc_AA800221_at - -
          Rat Genome U34 Set rc_AA924417_f_at - -
          Rat Genome U34 Set rc_AA924417_i_at - -
          Rat Genome U34 Set rc_AI171376_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21377859 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369003 - -
          RatRef-12 Expression BeadChip ILMN_53100 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0