Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593090_m1
          See other LOC100909664 GE Assays ›
          Gene Symbol:
          LOC100909664 Cep170
          Gene Name:
          centrosomal protein of 170 kDa-like
          centrosomal protein 170
          Gene Aliases:
          -
          KAB1, Kab, Rhk1
          Chromosome Location:
          Chr.13: 43806604 - 43845827 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 94807090 - 94887448 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00593090_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100909664
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751234.3 XP_003751282.2 6 - 7 1862 88
          XM_006249680.2 XP_006249742.1 6 - 7 1863 88
          XM_006249681.2 XP_006249743.1 6 - 7 1863 88
          XM_006249682.2 XP_006249744.1 6 - 7 1862 88
          XM_006249683.2 XP_006249745.1 6 - 7 1862 88
          Gene Symbol: Cep170
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008769812.1 XP_008768034.1 6 - 7 967 88
          GenBank mRNA AB038495.1 - 3 - 4 638 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P330612 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P330612 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P330612 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P330612 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19566 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5576 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27947 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770192 - -
          Affymetrix Rat Gene 1.0 ST Array 10770190 - -
          Affymetrix Rat Gene 1.0 ST Array 10770180 - -
          Affymetrix Rat Gene 1.0 ST Array 10770174 - -
          Affymetrix Rat Gene 1.0 ST Array 10770194 - -
          Affymetrix Rat Gene 1.0 ST Array 10770173 - -
          Affymetrix Rat Gene 1.0 ST Array 10770179 - -
          Affymetrix Rat Gene 1.0 ST Array 10770185 - -
          Affymetrix Rat Gene 1.0 ST Array 10770189 - -
          Affymetrix Rat Gene 1.0 ST Array 10770188 - -
          Affymetrix Rat Gene 1.0 ST Array 10770184 - -
          Affymetrix Rat Gene 1.0 ST Array 10770177 - -
          Affymetrix Rat Gene 1.0 ST Array 10770187 - -
          Affymetrix Rat Gene 1.0 ST Array 10770176 - -
          Affymetrix Rat Gene 1.0 ST Array 10770191 - -
          Affymetrix Rat Gene 1.0 ST Array 10770175 - -
          Affymetrix Rat Gene 1.0 ST Array 10770182 - -
          Affymetrix Rat Gene 1.0 ST Array 10770181 - -
          Affymetrix Rat Gene 1.0 ST Array 10770186 - -
          Affymetrix Rat Gene 1.0 ST Array 10770183 - -
          Affymetrix Rat Gene 1.0 ST Array 10770178 - -
          Affymetrix Rat Gene 1.0 ST Array 10770193 - -
          Affymetrix Rat Gene 1.1 ST Array 10770172 - -
          Rat Genome 230 2.0 Array 1387431_PM_at - -
          Rat Genome 230 2.0 Array 1387431_at - -
          Rat Genome 230 2.0 Array 1377664_PM_at - -
          Rat Genome 230 2.0 Array 1377664_at - -
          Affymetrix HT Rat Focus Array 1377664_at - -
          Affymetrix HT Rat Focus Array 1387431_PM_at - -
          Affymetrix HT Rat Focus Array 1387431_at - -
          Affymetrix HT Rat Focus Array 1377664_PM_at - -
          Rat Expression Set 230 1387431_at - -
          Rat Expression Set 230 1387431_PM_at - -
          Rat Expression Set 230 1377664_at - -
          Rat Expression Set 230 1377664_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770172 - -
          Rat Genome U34 Set rc_AI101205_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22178087 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-777976674b-tw4cl:80/100.66.134.228:80.
          git-commit: 6354fc3e62d97773410f28d6123c879be4bd0e1c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0