Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593424_m1
          See other KLC3 GE Assays ›
          Gene Symbol:
          Klc3
          Gene Name:
          kinesin light chain 3
          Gene Aliases:
          Klct
          Chromosome Location:
          Chr.1: 80306161 - 80315951 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593424_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138520.1 NP_612529.1 2 - 3 281 71
          XM_006228377.2 XP_006228439.1 2 - 3 434 71
          XM_006228378.2 XP_006228440.1 2 - 3 434 71
          XM_006228379.2 XP_006228441.1 2 - 3 356 71
          XM_006228380.2 XP_006228442.1 2 - 3 356 71
          GenBank mRNA AF166267.1 - 2 - 3 281 71
          BC078736.1 - 2 - 3 294 71
          BC097284.1 - 2 - 3 286 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004921.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klc3

          Gene Ontology Categories:

          Function(s) Process(es)

          axo-dendritic transport
          metabolic process
          microtubule motor activity
          microtubule binding
          kinesin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P334715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103887 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P334715 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P334715 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P334715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P334715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P334717 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700820860 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5472 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28832 - -
          Agilent Rat cDNA Microarray (G4105A) 700820860 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719483 - -
          Affymetrix Rat Gene 1.0 ST Array 10719484 - -
          Affymetrix Rat Gene 1.0 ST Array 10719482 - -
          Affymetrix Rat Gene 1.0 ST Array 10719479 - -
          Affymetrix Rat Gene 1.0 ST Array 10719478 - -
          Affymetrix Rat Gene 1.0 ST Array 10719481 - -
          Affymetrix Rat Gene 1.0 ST Array 10719485 - -
          Affymetrix Rat Gene 1.0 ST Array 10719474 - -
          Affymetrix Rat Gene 1.0 ST Array 10719476 - -
          Affymetrix Rat Gene 1.0 ST Array 10719480 - -
          Affymetrix Rat Gene 1.0 ST Array 10719475 - -
          Affymetrix Rat Gene 1.0 ST Array 10719477 - -
          Affymetrix Rat Gene 1.1 ST Array 10719473 - -
          Rat Genome 230 2.0 Array 1381537_at - -
          Rat Genome 230 2.0 Array 1388068_at - -
          Rat Genome 230 2.0 Array 1388068_PM_at - -
          Rat Genome 230 2.0 Array 1381537_PM_at - -
          Rat Genome 230 2.0 Array 1373735_at - -
          Rat Genome 230 2.0 Array 1373735_PM_at - -
          Affymetrix HT Rat Focus Array 1381537_PM_at - -
          Affymetrix HT Rat Focus Array 1388068_PM_at - -
          Affymetrix HT Rat Focus Array 1381537_at - -
          Affymetrix HT Rat Focus Array 1388068_at - -
          Affymetrix Rat Clariom S Assay PSR0100038814.rn.1 TC0100004921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100038810.rn.1 TC0100004921.rn.1 RTA TC
          Rat Expression Set 230 1388068_PM_at - -
          Rat Expression Set 230 1388068_at - -
          Rat Expression Set 230 1381537_PM_at - -
          Rat Expression Set 230 1381537_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719473 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21360850 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366127 - -
          RatRef-12 Expression BeadChip ILMN_59686 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0