Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597384_m1
          See other BDKRB2 GE Assays ›
          Gene Symbol:
          Bdkrb2
          Gene Name:
          bradykinin receptor B2
          Gene Aliases:
          B2BKR, B2BRA
          Chromosome Location:
          Chr.6: 129399468 - 129429676 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597384_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bdkrb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173100.2 NP_775123.2 2 - 3 209 75
          GenBank mRNA X69681.1 - 2 - 3 209 75

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the X69681.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus Rat Inflammation Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bdkrb2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          acute inflammatory response to antigenic stimulus
          smooth muscle contraction
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          negative regulation of cell proliferation
          response to salt stress
          sensory perception of pain
          negative regulation of peptidyl-serine phosphorylation
          maintenance of blood-brain barrier
          vasoconstriction
          vasodilation
          response to drug
          negative regulation of blood pressure
          arachidonic acid secretion
          cellular response to hypoxia
          negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
          negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress by p53 class mediator
          protease binding
          bradykinin receptor activity
          protein binding
          beta-2 adrenergic receptor binding
          type 1 angiotensin receptor binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25869 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036715 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036715 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P313516 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P758810 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6682 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886640 - -
          Affymetrix Rat Gene 1.0 ST Array 10886643 - -
          Affymetrix Rat Gene 1.0 ST Array 10886641 - -
          Affymetrix Rat Gene 1.0 ST Array 10886642 - -
          Affymetrix Rat Gene 1.0 ST Array 10886639 - -
          Affymetrix Rat Gene 1.1 ST Array 10886638 - -
          Rat Genome 230 2.0 Array 1370649_PM_at - -
          Rat Genome 230 2.0 Array 1370650_PM_s_at - -
          Rat Genome 230 2.0 Array 1370650_s_at - -
          Rat Genome 230 2.0 Array 1371120_PM_s_at - -
          Rat Genome 230 2.0 Array 1370649_at - -
          Rat Genome 230 2.0 Array 1371120_s_at - -
          Affymetrix HT Rat Focus Array 1371120_PM_s_at - -
          Affymetrix HT Rat Focus Array 1371120_s_at - -
          Affymetrix HT Rat Focus Array 1370649_at - -
          Affymetrix HT Rat Focus Array 1370650_s_at - -
          Affymetrix HT Rat Focus Array 1370650_PM_s_at - -
          Affymetrix HT Rat Focus Array 1370649_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600009925.rn.1 TC0600001429.rn.1 RTA TC
          Rat Expression Set 230 1370649_PM_at - -
          Rat Expression Set 230 1371120_s_at - -
          Rat Expression Set 230 1370650_s_at - -
          Rat Expression Set 230 1370649_at - -
          Rat Expression Set 230 1371120_PM_s_at - -
          Rat Expression Set 230 1370650_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886638 - -
          Rat Genome U34 Set X80187mRNA_s_at - -
          Rat Genome U34 Set X80187mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370361 - -
          RatRef-12 Expression BeadChip ILMN_63094 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix