Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00598533_m1
          See other PLOD2 GE Assays ›
          Gene Symbol:
          Plod2
          Gene Name:
          procollagen lysine, 2-oxoglutarate 5-dioxygenase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 99977334 - 100059736 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00598533_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plod2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142915.1 NP_001136387.1 4 - 5 697 67
          NM_175869.3 NP_787065.2 4 - 5 697 67
          GenBank mRNA AJ430860.1 - 4 - 5 759 67
          AJ430861.1 - 4 - 5 759 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001293.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plod2

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          cellular response to hormone stimulus
          molecular_function
          iron ion binding
          procollagen-lysine 5-dioxygenase activity
          L-ascorbic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119930 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90624 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006794 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006794 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006794 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409539 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P521151 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35646 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71048 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78383 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10912260 - -
          Affymetrix Rat Gene 1.0 ST Array 10912275 - -
          Affymetrix Rat Gene 1.0 ST Array 10912271 - -
          Affymetrix Rat Gene 1.0 ST Array 10912266 - -
          Affymetrix Rat Gene 1.0 ST Array 10912273 - -
          Affymetrix Rat Gene 1.0 ST Array 10912265 - -
          Affymetrix Rat Gene 1.0 ST Array 10912257 - -
          Affymetrix Rat Gene 1.0 ST Array 10912259 - -
          Affymetrix Rat Gene 1.0 ST Array 10912264 - -
          Affymetrix Rat Gene 1.0 ST Array 10912263 - -
          Affymetrix Rat Gene 1.0 ST Array 10912261 - -
          Affymetrix Rat Gene 1.0 ST Array 10912256 - -
          Affymetrix Rat Gene 1.0 ST Array 10912268 - -
          Affymetrix Rat Gene 1.0 ST Array 10912272 - -
          Affymetrix Rat Gene 1.0 ST Array 10912274 - -
          Affymetrix Rat Gene 1.0 ST Array 10912270 - -
          Affymetrix Rat Gene 1.0 ST Array 10912269 - -
          Affymetrix Rat Gene 1.0 ST Array 10912258 - -
          Affymetrix Rat Gene 1.0 ST Array 10912276 - -
          Affymetrix Rat Gene 1.0 ST Array 10912262 - -
          Affymetrix Rat Gene 1.0 ST Array 10912267 - -
          Affymetrix Rat Gene 1.1 ST Array 10912255 - -
          Rat Genome 230 2.0 Array 1372897_PM_at - -
          Rat Genome 230 2.0 Array 1372897_at - -
          Affymetrix HT Rat Focus Array 1372897_PM_at - -
          Affymetrix HT Rat Focus Array 1372897_at - -
          Affymetrix Rat Clariom S Assay PSR0800009462.rn.1 TC0800001293.rn.1 RTA TC
          Rat Expression Set 230 1372897_at - -
          Rat Expression Set 230 1372897_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10912255 - -
          Rat Genome U34 Set rc_AA892897_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21184803 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650259 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix