Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00683448_m1
          See other DUSP5 GE Assays ›
          Gene Symbol:
          Dusp5
          Gene Name:
          dual specificity phosphatase 5
          Gene Aliases:
          Cpg21
          Chromosome Location:
          Chr.1: 274245184 - 274258595 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00683448_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dusp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133578.1 NP_598262.1 3 - 4 922 63
          GenBank mRNA AF013144.1 - 3 - 4 922 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003823.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dusp5

          Gene Ontology Categories:

          Function(s) Process(es)

          endoderm formation
          protein dephosphorylation
          peptidyl-tyrosine dephosphorylation
          negative regulation of transcription, DNA-templated
          protein tyrosine phosphatase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P739860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17475 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P739860 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P739860 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P739860 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P739860 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77884 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37027 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716083 - -
          Affymetrix Rat Gene 1.0 ST Array 10716084 - -
          Affymetrix Rat Gene 1.0 ST Array 10716081 - -
          Affymetrix Rat Gene 1.0 ST Array 10716082 - -
          Affymetrix Rat Gene 1.1 ST Array 10716080 - -
          Rat Genome 230 2.0 Array 1368124_at - -
          Rat Genome 230 2.0 Array 1368124_PM_at - -
          Affymetrix HT Rat Focus Array 1368124_at - -
          Affymetrix HT Rat Focus Array 1368124_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100031173.rn.1 TC0100003823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100031175.rn.1 TC0100003823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100031174.rn.1 TC0100003823.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100031171.rn.1 TC0100003823.rn.1 RTA TC
          Rat Expression Set 230 1368124_PM_at - -
          Rat Expression Set 230 1368124_at - -
          Affymetrix Rat Gene 1.0 ST Array 10716080 - -
          Rat Genome U34 Set AF013144_at - -
          Rat Genome U34 Set AF013144_at - -
          Rat Genome U34 Set AF013144_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21946182 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355177 - -
          RatRef-12 Expression BeadChip ILMN_62284 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix