Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00689057_m1
          See other AMACR GE Assays ›
          Gene Symbol:
          Amacr
          Gene Name:
          alpha-methylacyl-CoA racemase
          Gene Aliases:
          Da1-8, Marc1
          Chromosome Location:
          Chr.2: 60949276 - 60961342 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00689057_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amacr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012816.2 NP_036948.2 4 - 5 740 99
          GenBank mRNA AY325250.1 - 989 99
          BC078885.1 - 4 - 5 708 99
          BC091163.1 - 4 - 5 709 99
          BC127505.1 - 4 - 5 729 99
          U89905.1 - 4 - 5 728 99
          Y08172.1 - 4 - 5 718 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000572.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Amacr

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid metabolic process
          bile acid biosynthetic process
          metabolic process
          bile acid metabolic process
          catalytic activity
          signaling receptor binding
          alpha-methylacyl-CoA racemase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P378885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6148 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P378885 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P378885 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P378885 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409654 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P460785 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21770 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68012 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813679 - -
          Affymetrix Rat Gene 1.0 ST Array 10813680 - -
          Affymetrix Rat Gene 1.0 ST Array 10813681 - -
          Affymetrix Rat Gene 1.0 ST Array 10813685 - -
          Affymetrix Rat Gene 1.0 ST Array 10813684 - -
          Affymetrix Rat Gene 1.1 ST Array 10813678 - -
          Rat Genome 230 2.0 Array 1367775_PM_at - -
          Rat Genome 230 2.0 Array 1367775_at - -
          Affymetrix HT Rat Focus Array 1367775_at - -
          Affymetrix HT Rat Focus Array 1367775_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200003593.rn.1 TC0200000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200003594.rn.1 TC0200000572.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200003595.rn.1 TC0200000572.rn.1 RTA TC
          Rat Expression Set 230 1367775_PM_at - -
          Rat Expression Set 230 1367775_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813678 - -
          Rat Genome U34 Set U89905_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21283472 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371494 - -
          RatRef-12 Expression BeadChip ILMN_53031 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline