Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00691721_g1
          See other MYH6 GE Assays ›
          Gene Symbol:
          Myh6
          Gene Name:
          myosin, heavy chain 6, cardiac muscle, alpha
          Gene Aliases:
          Myhca
          Chromosome Location:
          Chr.15: 33605653 - 33629592 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00691721_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myh6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017239.2 NP_058935.2 9 - 10 932 71
          GenBank mRNA X15938.1 - 9 - 10 929 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001688.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myh6

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          regulation of the force of heart contraction
          regulation of heart rate
          muscle contraction
          striated muscle contraction
          adult heart development
          visceral muscle development
          regulation of heart contraction
          metabolic process
          regulation of blood pressure
          response to heat
          actin filament-based movement
          muscle filament sliding
          myofibril assembly
          BMP signaling pathway
          regulation of ATPase activity
          sarcomere organization
          cardiac muscle fiber development
          atrial cardiac muscle tissue morphogenesis
          ventricular cardiac muscle tissue morphogenesis
          canonical Wnt signaling pathway
          regulation of heart growth
          microfilament motor activity
          actin binding
          calmodulin binding
          ATP binding
          ATPase activity
          protein kinase binding
          actin-dependent ATPase activity
          calcium-dependent ATPase activity
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P756334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923057 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107651 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P756334 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P756334 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P756334 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P756334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102693 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44504 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27315 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783697 - -
          Affymetrix Rat Gene 1.0 ST Array 10783705 - -
          Affymetrix Rat Gene 1.0 ST Array 10783699 - -
          Affymetrix Rat Gene 1.0 ST Array 10783712 - -
          Affymetrix Rat Gene 1.0 ST Array 10783703 - -
          Affymetrix Rat Gene 1.0 ST Array 10783701 - -
          Affymetrix Rat Gene 1.0 ST Array 10783688 - -
          Affymetrix Rat Gene 1.0 ST Array 10783692 - -
          Affymetrix Rat Gene 1.0 ST Array 10783696 - -
          Affymetrix Rat Gene 1.0 ST Array 10783708 - -
          Affymetrix Rat Gene 1.0 ST Array 10783698 - -
          Affymetrix Rat Gene 1.0 ST Array 10783695 - -
          Affymetrix Rat Gene 1.0 ST Array 10783706 - -
          Affymetrix Rat Gene 1.0 ST Array 10783702 - -
          Affymetrix Rat Gene 1.0 ST Array 10783700 - -
          Affymetrix Rat Gene 1.0 ST Array 10783694 - -
          Affymetrix Rat Gene 1.0 ST Array 10783690 - -
          Affymetrix Rat Gene 1.0 ST Array 10783710 - -
          Affymetrix Rat Gene 1.0 ST Array 10783709 - -
          Affymetrix Rat Gene 1.0 ST Array 10783691 - -
          Affymetrix Rat Gene 1.0 ST Array 10783693 - -
          Affymetrix Rat Gene 1.0 ST Array 10783707 - -
          Affymetrix Rat Gene 1.0 ST Array 10783687 - -
          Affymetrix Rat Gene 1.0 ST Array 10783689 - -
          Affymetrix Rat Gene 1.0 ST Array 10783711 - -
          Affymetrix Rat Gene 1.0 ST Array 10783704 - -
          Affymetrix Rat Gene 1.1 ST Array 10783686 - -
          Rat Genome 230 2.0 Array 1367928_PM_at - -
          Rat Genome 230 2.0 Array 1387049_PM_at - -
          Rat Genome 230 2.0 Array 1368093_at - -
          Rat Genome 230 2.0 Array 1396165_PM_at - -
          Rat Genome 230 2.0 Array 1398248_s_at - -
          Rat Genome 230 2.0 Array 1387049_at - -
          Rat Genome 230 2.0 Array 1398248_PM_s_at - -
          Rat Genome 230 2.0 Array 1367928_at - -
          Rat Genome 230 2.0 Array 1368093_PM_at - -
          Rat Genome 230 2.0 Array 1396165_at - -
          Affymetrix HT Rat Focus Array 1396165_PM_at - -
          Affymetrix HT Rat Focus Array 1387049_at - -
          Affymetrix HT Rat Focus Array 1387049_PM_at - -
          Affymetrix HT Rat Focus Array 1396165_at - -
          Affymetrix HT Rat Focus Array 1368093_PM_at - -
          Affymetrix HT Rat Focus Array 1368093_at - -
          Affymetrix Rat Clariom S Assay PSR1500010837.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010852.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010810.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010839.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010822.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010831.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010812.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010856.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010826.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010814.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010825.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010816.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010861.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010813.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010853.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010846.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010851.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010832.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010860.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010836.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010815.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010858.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010863.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010819.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010850.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010838.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010859.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010848.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010844.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010820.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010830.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010840.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010834.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010862.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010855.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010833.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010847.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010857.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010818.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010849.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010854.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010845.rn.1 TC1500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010842.rn.1 TC1500001688.rn.1 RTA TC
          Rat Expression Set 230 1387049_PM_at - -
          Rat Expression Set 230 1368093_PM_at - -
          Rat Expression Set 230 1398248_PM_s_at - -
          Rat Expression Set 230 1368093_at - -
          Rat Expression Set 230 1387049_at - -
          Rat Expression Set 230 1398248_s_at - -
          Rat Expression Set 230 1396165_at - -
          Rat Expression Set 230 1396165_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783686 - -
          Rat Genome U34 Set rc_AI103920_f_at - -
          Rat Genome U34 Set X15939_f_at - -
          Rat Genome U34 Set rc_AI104924_f_at - -
          Rat Genome U34 Set X15939_r_at - -
          Rat Genome U34 Set rc_AA891522_f_at - -
          Rat Genome U34 Set rc_AI009388_at - -
          Rat Genome U34 Set rc_AI232024_f_at - -
          Rat Genome U34 Set rc_AA819464_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21928845 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349554 - -
          RatRef-12 Expression BeadChip ILMN_69653 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0