Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755291_g1
          See other PSMA5 GE Assays ›
          Gene Symbol:
          Psma5
          Gene Name:
          proteasome (prosome, macropain) subunit, alpha type 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 211050385 - 211073703 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755291_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psma5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017282.1 NP_058978.1 5 - 6 482 69
          GenBank mRNA BC060575.1 - 5 - 6 503 69
          D10756.1 - 5 - 6 482 69
          FQ218317.1 - 5 - 6 499 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001986.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psma5

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          threonine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P721011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33429 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139884 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P721011 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P721011 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P721011 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P721011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103661 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10951 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818320 - -
          Affymetrix Rat Gene 1.0 ST Array 10818322 - -
          Affymetrix Rat Gene 1.0 ST Array 10818323 - -
          Affymetrix Rat Gene 1.0 ST Array 10818321 - -
          Affymetrix Rat Gene 1.0 ST Array 10818318 - -
          Affymetrix Rat Gene 1.0 ST Array 10818319 - -
          Affymetrix Rat Gene 1.0 ST Array 10818324 - -
          Affymetrix Rat Gene 1.0 ST Array 10818325 - -
          Affymetrix Rat Gene 1.1 ST Array 10818317 - -
          Rat Genome 230 2.0 Array 1387884_PM_at - -
          Rat Genome 230 2.0 Array 1387884_at - -
          Affymetrix HT Rat Focus Array 1387884_at - -
          Affymetrix HT Rat Focus Array 1387884_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200013644.rn.1 TC0200001986.rn.1 RTA TC
          Rat Expression Set 230 1387884_at - -
          Rat Expression Set 230 1387884_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818317 - -
          Rat Genome U34 Set rc_AA891383_i_at - -
          Rat Genome U34 Set D10756_g_at - -
          Rat Genome U34 Set D10756_at - -
          Rat Genome U34 Set rc_AI169479_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21710432 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355230 - -
          RatRef-12 Expression BeadChip ILMN_49549 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix