Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00788171_m1
          See other STRADA GE Assays ›
          Gene Symbol:
          Strada
          Gene Name:
          STE20-related kinase adaptor alpha
          Gene Aliases:
          Lyk5
          Chromosome Location:
          Chr.10: 94355369 - 94384404 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00788171_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Strada
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182820.1 NP_877972.1 8 - 9 867 67
          XM_006247586.2 XP_006247648.1 10 - 11 1046 67
          XM_006247588.2 XP_006247650.1 10 - 11 1045 67
          XM_006247595.2 XP_006247657.1 9 - 10 1015 67
          XM_008768359.1 XP_008766581.1 10 - 11 1046 67
          XM_008768360.1 XP_008766582.1 10 - 11 1022 67
          XM_008768361.1 XP_008766583.1 8 - 9 958 67
          XM_008768362.1 XP_008766584.1 8 - 9 934 67
          XM_008768363.1 XP_008766585.1 9 - 10 1016 67
          XM_008768364.1 XP_008766586.1 9 - 10 992 67
          GenBank mRNA AY327409.1 - 8 - 9 867 67
          BC081911.1 - 9 - 10 951 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004419.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Strada

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          protein phosphorylation
          protein export from nucleus
          activation of protein kinase activity
          intracellular signal transduction
          protein heterooligomerization
          positive regulation of protein serine/threonine kinase activity
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase binding
          protein kinase activator activity
          protein serine/threonine kinase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10545 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217471 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74894 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10545 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10545 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49904 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55852 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59944 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748210 - -
          Affymetrix Rat Gene 1.0 ST Array 10748206 - -
          Affymetrix Rat Gene 1.0 ST Array 10748220 - -
          Affymetrix Rat Gene 1.0 ST Array 10748219 - -
          Affymetrix Rat Gene 1.0 ST Array 10748215 - -
          Affymetrix Rat Gene 1.0 ST Array 10748209 - -
          Affymetrix Rat Gene 1.0 ST Array 10748211 - -
          Affymetrix Rat Gene 1.0 ST Array 10748213 - -
          Affymetrix Rat Gene 1.0 ST Array 10748212 - -
          Affymetrix Rat Gene 1.0 ST Array 10748218 - -
          Affymetrix Rat Gene 1.0 ST Array 10748207 - -
          Affymetrix Rat Gene 1.0 ST Array 10748216 - -
          Affymetrix Rat Gene 1.0 ST Array 10748214 - -
          Affymetrix Rat Gene 1.1 ST Array 10748205 - -
          Rat Genome 230 2.0 Array 1398400_PM_at - -
          Rat Genome 230 2.0 Array 1398400_at - -
          Affymetrix HT Rat Focus Array 1398400_at - -
          Affymetrix HT Rat Focus Array 1398400_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000041638.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041621.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041625.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041630.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041616.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041617.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041622.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041628.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041631.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041642.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041624.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041627.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041629.rn.1 TC1000004419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041636.rn.1 TC1000004419.rn.1 RTA TC
          Rat Expression Set 230 1398400_PM_at - -
          Rat Expression Set 230 1398400_at - -
          Affymetrix Rat Gene 1.0 ST Array 10748205 - -
          Rat Genome U34 Set rc_AA963467_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20780499 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362000 - -
          RatRef-12 Expression BeadChip ILMN_65907 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0