Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821690_g1
          See other ATP6V0C GE Assays ›
          Gene Symbol:
          Atp6v0c
          Gene Name:
          ATPase, H+ transporting, lysosomal V0 subunit C
          Gene Aliases:
          Atp6c, Atp6l
          Chromosome Location:
          Chr.10: 13537031 - 13543407 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821690_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp6v0c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130823.3 NP_570836.1 2 - 3 418 67
          XM_006245898.1 XP_006245960.1 3 - 4 919 67
          GenBank mRNA BC063154.1 - 2 - 3 422 67
          D10874.1 - 2 - 3 359 67
          FQ211954.1 - 2 - 3 398 67
          FQ212559.1 - 2 - 3 402 67
          FQ212637.1 - 2 - 3 398 67
          FQ212733.1 - 2 - 3 402 67
          FQ213105.1 - 2 - 3 402 67
          FQ213455.1 - 2 - 3 402 67
          FQ214388.1 - 2 - 3 402 67
          FQ214503.1 - 2 - 3 423 67
          FQ216440.1 - 2 - 3 402 67
          FQ220211.1 - 2 - 3 402 67
          FQ220254.1 - 2 - 3 402 67
          FQ220713.1 - 2 - 3 423 67
          FQ220906.1 - 2 - 3 371 67
          FQ221569.1 - 2 - 3 413 67
          FQ229453.1 - 2 - 3 398 67
          FQ229871.1 - 2 - 3 403 67
          FQ234225.1 - 2 - 3 423 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp6v0c

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of Wnt signaling pathway
          proton transmembrane transporter activity
          ubiquitin protein ligase binding
          ATPase-coupled ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P440280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289551 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88318 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P440280 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P440280 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P440280 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10171 - -
          Agilent Rat cDNA Microarray (G4105A) 700544301 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740997 - -
          Affymetrix Rat Gene 1.0 ST Array 10741000 - -
          Affymetrix Rat Gene 1.0 ST Array 10740999 - -
          Affymetrix Rat Gene 1.0 ST Array 10740998 - -
          Affymetrix Rat Gene 1.1 ST Array 10740996 - -
          Rat Genome 230 2.0 Array 1398755_PM_at - -
          Rat Genome 230 2.0 Array 1398755_at - -
          Affymetrix HT Rat Focus Array 1398755_at - -
          Affymetrix HT Rat Focus Array 1398755_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000021021.rn.1 TC1000002380.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021023.rn.1 TC1000002380.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000021022.rn.1 TC1000002380.rn.1 RTA TC
          Rat Expression Set 230 1398755_PM_at - -
          Rat Expression Set 230 1398755_at - -
          Affymetrix Rat Gene 1.0 ST Array 10740996 - -
          Rat Genome U34 Set D10874_g_at - -
          Rat Genome U34 Set D10874_at - -
          Rat Genome U34 Set rc_AI177126_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21177319 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363169 - -
          RatRef-12 Expression BeadChip ILMN_53801 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0