Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821697_g1
          See other STK17B GE Assays ›
          Gene Symbol:
          Stk17b
          Gene Name:
          serine/threonine kinase 17b
          Gene Aliases:
          Drak2
          Chromosome Location:
          Chr.9: 60399261 - 60430081 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821697_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stk17b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133392.1 NP_596883.1 5 - 6 775 79
          XM_006244908.2 XP_006244970.1 5 - 6 1544 79
          GenBank mRNA AB070349.1 - 5 - 6 775 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stk17b

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          apoptotic process
          programmed cell death
          intracellular signal transduction
          protein autophosphorylation
          positive regulation of fibroblast apoptotic process
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148321 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19672 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13172 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13172 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13172 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22552 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14302 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64576 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10927995 - -
          Affymetrix Rat Gene 1.0 ST Array 10927996 - -
          Affymetrix Rat Gene 1.0 ST Array 10927998 - -
          Affymetrix Rat Gene 1.0 ST Array 10927997 - -
          Affymetrix Rat Gene 1.0 ST Array 10928000 - -
          Affymetrix Rat Gene 1.0 ST Array 10927994 - -
          Affymetrix Rat Gene 1.0 ST Array 10927993 - -
          Affymetrix Rat Gene 1.0 ST Array 10927999 - -
          Affymetrix Rat Gene 1.1 ST Array 10927992 - -
          Rat Genome 230 2.0 Array 1373164_PM_at - -
          Rat Genome 230 2.0 Array 1393956_at - -
          Rat Genome 230 2.0 Array 1387502_PM_at - -
          Rat Genome 230 2.0 Array 1387502_at - -
          Rat Genome 230 2.0 Array 1373164_at - -
          Rat Genome 230 2.0 Array 1393956_PM_at - -
          Affymetrix HT Rat Focus Array 1387502_PM_at - -
          Affymetrix HT Rat Focus Array 1387502_at - -
          Affymetrix Rat Clariom S Assay PSR0900016184.rn.1 TC0900001972.rn.1 RTA TC
          Rat Expression Set 230 1387502_at - -
          Rat Expression Set 230 1387502_PM_at - -
          Rat Expression Set 230 1393956_at - -
          Rat Expression Set 230 1393956_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10927992 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21058915 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370817 - -
          RatRef-12 Expression BeadChip ILMN_49397 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix