Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00822054_g1
          See other ISCA1 GE Assays ›
          Gene Symbol:
          Isca1
          Gene Name:
          iron-sulfur cluster assembly 1
          Gene Aliases:
          Hbld2, hIscA
          Chromosome Location:
          Chr.17: 5281727 - 5294386 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00822054_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Isca1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181626.3 NP_853657.1 1 - 2 136 98
          GenBank mRNA AF508158.1 - 1 - 2 102 98
          BC070929.1 - 1 - 2 136 98
          BC078677.1 - 1 - 2 146 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000070.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Isca1

          Gene Ontology Categories:

          Function(s) Process(es)

          iron-sulfur cluster assembly
          endocrine process
          structural molecule activity
          metal ion binding
          iron-sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022751 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P367726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P367726 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022751 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022751 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P367726 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P367726 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022751 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P367726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10793736 - -
          Affymetrix Rat Gene 1.0 ST Array 10793735 - -
          Affymetrix Rat Gene 1.0 ST Array 10793737 - -
          Affymetrix Rat Gene 1.0 ST Array 10793739 - -
          Affymetrix Rat Gene 1.0 ST Array 10793740 - -
          Affymetrix Rat Gene 1.0 ST Array 10871443 - -
          Affymetrix Rat Gene 1.0 ST Array 10793738 - -
          Affymetrix Rat Gene 1.0 ST Array 10787020 - -
          Affymetrix Rat Gene 1.1 ST Array 10871435 - -
          Affymetrix Rat Gene 1.1 ST Array 10793734 - -
          Affymetrix Rat Gene 1.1 ST Array 10787019 - -
          Rat Genome 230 2.0 Array 1388613_PM_at - -
          Rat Genome 230 2.0 Array 1388613_at - -
          Rat Genome 230 2.0 Array 1388603_PM_a_at - -
          Rat Genome 230 2.0 Array 1388603_a_at - -
          Affymetrix HT Rat Focus Array 1388613_at - -
          Affymetrix HT Rat Focus Array 1388603_a_at - -
          Affymetrix HT Rat Focus Array 1388603_PM_a_at - -
          Affymetrix HT Rat Focus Array 1388613_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700000497.rn.1 TC1700000070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000493.rn.1 TC1700000070.rn.1 RTA TC
          Rat Expression Set 230 1388603_PM_a_at - -
          Rat Expression Set 230 1388613_PM_at - -
          Rat Expression Set 230 1388613_at - -
          Rat Expression Set 230 1388603_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10793734 - -
          Affymetrix Rat Gene 1.0 ST Array 10871435 - -
          Affymetrix Rat Gene 1.0 ST Array 10787019 - -
          Rat Genome U34 Set rc_AA946469_at - -
          Rat Genome U34 Set rc_AI231572_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21419639 - -
          ABI Rat Genome Survey Microarray 21773650 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371925 - -
          RatRef-12 Expression BeadChip ILMN_57967 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0