Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01074556_m1
          See other ENY2 GE Assays ›
          Gene Symbol:
          Eny2
          Gene Name:
          enhancer of yellow 2 homolog (Drosophila)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 83348656 - 83358430 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01074556_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eny2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130580.1 NP_001124052.1 4 - 5 357 67
          XM_006241615.2 XP_006241677.1 4 - 5 323 67
          GenBank mRNA FQ223243.1 - 4 - 5 241 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001063.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription elongation from RNA polymerase II promoter
          mRNA export from nucleus
          protein transport
          histone deubiquitination
          positive regulation of transcription, DNA-templated
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          molecular_function
          transcription coactivator activity
          nuclear receptor coactivator activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P256181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540598 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P256181 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P256181 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P256181 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P220541 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P256181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896403 - -
          Affymetrix Rat Gene 1.0 ST Array 10896401 - -
          Affymetrix Rat Gene 1.0 ST Array 10896404 - -
          Affymetrix Rat Gene 1.0 ST Array 10896402 - -
          Affymetrix Rat Gene 1.1 ST Array 10896400 - -
          Rat Genome 230 2.0 Array 1376606_PM_a_at - -
          Rat Genome 230 2.0 Array 1376606_a_at - -
          Affymetrix HT Rat Focus Array 1376606_PM_a_at - -
          Affymetrix HT Rat Focus Array 1376606_a_at - -
          Affymetrix Rat Clariom S Assay PSR0700007917.rn.1 TC0700001063.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700007923.rn.1 TC0700001063.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700007920.rn.1 TC0700001063.rn.1 RTA TC
          Rat Expression Set 230 1376606_a_at - -
          Rat Expression Set 230 1376606_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896400 - -
          Rat Genome U34 Set rc_AI169290_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22186445 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0