Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01236170_g1
          See other COX8A GE Assays ›
          Gene Symbol:
          Cox8a
          Gene Name:
          cytochrome c oxidase subunit VIIIa
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 222466575 - 222468896 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01236170_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox8a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134345.1 NP_599172.1 1 - 2 171 83
          GenBank mRNA BC126066.1 - 1 - 2 138 83
          CB701133.1 - 1 - 2 182 83
          L48209.1 - 1 - 2 89 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007192.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox8a

          Gene Ontology Categories:

          Function(s) Process(es)

          proton transmembrane transport
          cytochrome-c oxidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P570756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228095 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P570756 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P570756 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P570756 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P570756 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728384 - -
          Affymetrix Rat Gene 1.0 ST Array 10728385 - -
          Affymetrix Rat Gene 1.1 ST Array 10728383 - -
          Rat Genome 230 2.0 Array 1388113_at - -
          Rat Genome 230 2.0 Array 1388113_PM_at - -
          Affymetrix HT Rat Focus Array 1388113_at - -
          Affymetrix HT Rat Focus Array 1388113_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100057933.rn.1 TC0100007192.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057934.rn.1 TC0100007192.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057937.rn.1 TC0100007192.rn.1 RTA TC
          Rat Expression Set 230 1388113_PM_at - -
          Rat Expression Set 230 1388113_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728383 - -
          Rat Genome U34 Set rc_AI102505_g_at - -
          Rat Genome U34 Set L48209_s_at - -
          Rat Genome U34 Set M28255_s_at - -
          Rat Genome U34 Set rc_AI102505_at - -
          Rat Genome U34 Set rc_AI102505_at - -
          Rat Genome U34 Set rc_AI102505_g_at - -
          Rat Genome U34 Set L48209_s_at - -
          Rat Genome U34 Set M28255_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369379 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0