Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012749.2 | NP_036881.2 | 7 - 8 | 1311 | 131 | |
GenBank mRNA | BC085751.1 | - | 7 - 8 | 1311 | 131 | |
FJ817497.1 | - | 7 - 8 | 750 | 131 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |