Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400908_m1
          See other LOC100366216 GE Assays ›
          Gene Symbol:
          LOC100366216 LOC103690030 Sp100
          Gene Name:
          nuclear antigen Sp100-like
          nuclear autoantigen Sp-100-like
          SP100 nuclear antigen
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.9: 90964841 - 90985313 on Build Rnor_6.0
          Chromosome Location:
          Chr.9: 90993702 - 91011649 on Build Rnor_6.0
          Chromosome Location:
          Chr.9: 92675103 - 92718489 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn01400908_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100366216
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750725.3 XP_003750773.1 2 - 3 1348 71
          XM_006245340.2 XP_006245402.1 3 - 4 2010 71
          XM_006245341.1 XP_006245403.1 3 - 4 279 71
          XM_006245342.2 XP_006245404.1 3 - 4 2009 71
          XM_006245343.2 XP_006245405.1 2 - 3 1351 71
          XM_006245344.2 XP_006245406.1 2 - 3 581 71
          Gene Symbol: LOC103690030
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006245338.2 XP_006245400.1 2 - 3 287 71
          XM_006245339.2 XP_006245401.1 2 - 3 761 71
          XM_008767326.1 XP_008765548.1 1 - 2 329 71
          XM_008767327.1 XP_008765549.1 2 - 3 287 71
          Gene Symbol: Sp100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014220.1 NP_001014242.1 1 - 2 180 71
          GenBank mRNA BC083808.1 - 1 - 2 180 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002707.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100366216
          Click here to find siRNAs for Gene LOC103690030
          Click here to find siRNAs for Gene Sp100

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          telomere maintenance
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          negative regulation of endothelial cell migration
          response to retinoic acid
          negative regulation of viral transcription
          response to cytokine
          response to type I interferon
          response to interferon-gamma
          negative regulation of DNA binding
          negative regulation of DNA-binding transcription factor activity
          regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of protein export from nucleus
          positive regulation of DNA-binding transcription factor activity
          negative regulation of cellular component movement
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of Fas signaling pathway
          negative regulation of nucleic acid-templated transcription
          transcription coactivator activity
          transcription corepressor activity
          transcription factor binding
          kinase binding
          protein domain specific binding
          identical protein binding
          protein homodimerization activity
          chromo shadow domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0900022182.rn.1 TC0900002707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022183.rn.1 TC0900002707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022184.rn.1 TC0900002707.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900022186.rn.1 TC0900002707.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0