Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402412_m1
          See other TTC33 GE Assays ›
          Gene Symbol:
          Ttc33
          Gene Name:
          tetratricopeptide repeat domain 33
          Gene Aliases:
          RGD1564042
          Chromosome Location:
          Chr.2: 54897363 - 54944241 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402412_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttc33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106414.1 NP_001099884.1 4 - 5 631 65
          XM_006231993.2 XP_006232055.1 4 - 5 520 65
          XM_006231994.1 XP_006232056.1 4 - 5 501 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000493.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796359 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140963 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130061 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17779 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17779 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17779 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813352 - -
          Affymetrix Rat Gene 1.0 ST Array 10813349 - -
          Affymetrix Rat Gene 1.0 ST Array 10813350 - -
          Affymetrix Rat Gene 1.0 ST Array 10813351 - -
          Affymetrix Rat Gene 1.0 ST Array 10813348 - -
          Affymetrix Rat Gene 1.1 ST Array 10813347 - -
          Rat Genome 230 2.0 Array 1390793_PM_at - -
          Rat Genome 230 2.0 Array 1390793_at - -
          Affymetrix HT Rat Focus Array 1390793_at - -
          Affymetrix HT Rat Focus Array 1390793_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200002953.rn.1 TC0200000493.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002952.rn.1 TC0200000493.rn.1 RTA TC
          Rat Expression Set 230 1390793_at - -
          Rat Expression Set 230 1390793_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813347 - -
          Rat Genome U34 Set rc_AA850551_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20920041 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357874 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix