Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402867_m1
          See other TLL1 GE Assays ›
          Gene Symbol:
          Tll1
          Gene Name:
          tolloid-like 1
          Gene Aliases:
          Tll
          Chromosome Location:
          Chr.16: 27399467 - 27597240 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402867_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tll1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106081.1 NP_001099551.1 12 - 13 2137 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000398.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          metalloendopeptidase activity
          calcium ion binding
          collagen binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P323700 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146585 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93523 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164377 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109707 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111016 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P323700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105565 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P323700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105565 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P323700 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105565 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3731 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92073 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58204 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 235 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10787857 - -
          Affymetrix Rat Gene 1.0 ST Array 10787873 - -
          Affymetrix Rat Gene 1.0 ST Array 10787875 - -
          Affymetrix Rat Gene 1.0 ST Array 10787863 - -
          Affymetrix Rat Gene 1.0 ST Array 10787862 - -
          Affymetrix Rat Gene 1.0 ST Array 10787865 - -
          Affymetrix Rat Gene 1.0 ST Array 10787868 - -
          Affymetrix Rat Gene 1.0 ST Array 10787870 - -
          Affymetrix Rat Gene 1.0 ST Array 10787860 - -
          Affymetrix Rat Gene 1.0 ST Array 10787869 - -
          Affymetrix Rat Gene 1.0 ST Array 10787861 - -
          Affymetrix Rat Gene 1.0 ST Array 10787867 - -
          Affymetrix Rat Gene 1.0 ST Array 10787858 - -
          Affymetrix Rat Gene 1.0 ST Array 10787872 - -
          Affymetrix Rat Gene 1.0 ST Array 10787876 - -
          Affymetrix Rat Gene 1.0 ST Array 10787871 - -
          Affymetrix Rat Gene 1.0 ST Array 10787864 - -
          Affymetrix Rat Gene 1.0 ST Array 10787877 - -
          Affymetrix Rat Gene 1.0 ST Array 10787859 - -
          Affymetrix Rat Gene 1.0 ST Array 10787866 - -
          Affymetrix Rat Gene 1.0 ST Array 10787874 - -
          Affymetrix Rat Gene 1.1 ST Array 10787856 - -
          Affymetrix Rat Clariom S Assay PSR1600003811.rn.1 TC1600000398.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10787856 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix