Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01405725_m1
          See other LOC103689930 GE Assays ›
          Gene Symbol:
          LOC103689930 Trappc8
          Gene Name:
          trafficking protein particle complex subunit 8
          trafficking protein particle complex 8
          Gene Aliases:
          -
          RGD1309102
          Chromosome Location:
          Chr.18: 15298678 - 15375034 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 12070081 - 12116697 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01405725_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103689930
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006254461.2 XP_006254523.1 28 - 29 4341 115
          XM_006254462.2 XP_006254524.1 28 - 29 4338 115
          XM_008771994.1 XP_008770216.1 28 - 29 4395 115
          Gene Symbol: Trappc8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106160.1 NP_001099630.1 4095 115
          GenBank mRNA BC166545.1 - 150 115
          FQ230517.1 - 1952 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000136.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          autophagosome assembly
          endoplasmic reticulum to Golgi vesicle-mediated transport
          autophagy of peroxisome
          cytoplasm to vacuole transport by the Cvt pathway
          protein localization to phagophore assembly site
          autophagy of nucleus

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P472472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49170 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108726 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P472472 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P472472 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P472472 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P529905 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101776 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803416 - -
          Affymetrix Rat Gene 1.0 ST Array 10803433 - -
          Affymetrix Rat Gene 1.0 ST Array 10803428 - -
          Affymetrix Rat Gene 1.0 ST Array 10803410 - -
          Affymetrix Rat Gene 1.0 ST Array 10803425 - -
          Affymetrix Rat Gene 1.0 ST Array 10803427 - -
          Affymetrix Rat Gene 1.0 ST Array 10803408 - -
          Affymetrix Rat Gene 1.0 ST Array 10803430 - -
          Affymetrix Rat Gene 1.0 ST Array 10803420 - -
          Affymetrix Rat Gene 1.0 ST Array 10803411 - -
          Affymetrix Rat Gene 1.0 ST Array 10803413 - -
          Affymetrix Rat Gene 1.0 ST Array 10803418 - -
          Affymetrix Rat Gene 1.0 ST Array 10803421 - -
          Affymetrix Rat Gene 1.0 ST Array 10803419 - -
          Affymetrix Rat Gene 1.0 ST Array 10803415 - -
          Affymetrix Rat Gene 1.0 ST Array 10803407 - -
          Affymetrix Rat Gene 1.0 ST Array 10803426 - -
          Affymetrix Rat Gene 1.0 ST Array 10803417 - -
          Affymetrix Rat Gene 1.0 ST Array 10803423 - -
          Affymetrix Rat Gene 1.0 ST Array 10803414 - -
          Affymetrix Rat Gene 1.0 ST Array 10803429 - -
          Affymetrix Rat Gene 1.0 ST Array 10803422 - -
          Affymetrix Rat Gene 1.0 ST Array 10803431 - -
          Affymetrix Rat Gene 1.0 ST Array 10803406 - -
          Affymetrix Rat Gene 1.0 ST Array 10803424 - -
          Affymetrix Rat Gene 1.0 ST Array 10803412 - -
          Affymetrix Rat Gene 1.0 ST Array 10803409 - -
          Affymetrix Rat Gene 1.1 ST Array 10803405 - -
          Rat Genome 230 2.0 Array 1376609_PM_at - -
          Rat Genome 230 2.0 Array 1376609_at - -
          Affymetrix HT Rat Focus Array 1376609_PM_at - -
          Affymetrix HT Rat Focus Array 1376609_at - -
          Affymetrix Rat Clariom S Assay PSR1800000958.rn.1 TC1800000136.rn.1 RTA TC
          Rat Expression Set 230 1376609_at - -
          Rat Expression Set 230 1376609_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803405 - -
          Rat Genome U34 Set rc_AA901020_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20768488 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352891 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0