Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01407078_m1
          See other SH3BGR GE Assays ›
          Gene Symbol:
          Sh3bgr
          Gene Name:
          SH3 domain binding glutamate-rich protein
          Gene Aliases:
          RGD1563599
          Chromosome Location:
          Chr.11: 36555354 - 36579537 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01407078_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sh3bgr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127682.1 NP_001121154.1 6 - 7 736 132
          XM_006248184.2 XP_006248246.1 7 - 8 852 132
          GenBank mRNA BC159440.1 - 7 - 8 754 132
          FQ215323.1 - 7 - 8 758 132
          FQ216183.1 - 7 - 8 657 132
          FQ216585.1 - 7 - 8 655 132
          FQ216690.1 - 7 - 8 707 132
          FQ223676.1 - 7 - 8 708 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000353.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P471450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78134 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P471450 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P471450 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P471450 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P471450 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6766 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750480 - -
          Affymetrix Rat Gene 1.0 ST Array 10750481 - -
          Affymetrix Rat Gene 1.0 ST Array 10750482 - -
          Affymetrix Rat Gene 1.0 ST Array 10750485 - -
          Affymetrix Rat Gene 1.0 ST Array 10750488 - -
          Affymetrix Rat Gene 1.0 ST Array 10750483 - -
          Affymetrix Rat Gene 1.0 ST Array 10750487 - -
          Affymetrix Rat Gene 1.0 ST Array 10750484 - -
          Affymetrix Rat Gene 1.1 ST Array 10750479 - -
          Rat Genome 230 2.0 Array 1372296_at - -
          Rat Genome 230 2.0 Array 1372296_PM_at - -
          Affymetrix HT Rat Focus Array 1372296_at - -
          Affymetrix HT Rat Focus Array 1372296_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100001731.rn.1 TC1100000353.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001729.rn.1 TC1100000353.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001730.rn.1 TC1100000353.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001728.rn.1 TC1100000353.rn.1 RTA TC
          Rat Expression Set 230 1372296_at - -
          Rat Expression Set 230 1372296_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750479 - -
          Rat Genome U34 Set rc_AI044101_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21520404 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363643 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0