Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01409756_m1
          See other LOC100365773 GE Assays ›
          Gene Symbol:
          LOC100365773
          Gene Name:
          pink-eyed dilution-like
          Gene Aliases:
          Oca2, p
          Chromosome Location:
          Chr.1: 114661970 - 114987433 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01409756_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100365773
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271493.1 NP_001258422.1 22 - 23 2475 81
          GenBank mRNA AB191343.1 - 22 - 23 2450 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001608.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          biological_process
          cell population proliferation
          melanocyte differentiation
          melanin biosynthetic process
          pigmentation
          developmental pigmentation
          transmembrane transport
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P332890 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P332890 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71861 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38792 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75940 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69624 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80139 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89574 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49444 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104024 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71254 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10707566 - -
          Affymetrix Rat Gene 1.0 ST Array 10707570 - -
          Affymetrix Rat Gene 1.0 ST Array 10707571 - -
          Affymetrix Rat Gene 1.0 ST Array 10707558 - -
          Affymetrix Rat Gene 1.0 ST Array 10707564 - -
          Affymetrix Rat Gene 1.0 ST Array 10707565 - -
          Affymetrix Rat Gene 1.0 ST Array 10707551 - -
          Affymetrix Rat Gene 1.0 ST Array 10707549 - -
          Affymetrix Rat Gene 1.0 ST Array 10707554 - -
          Affymetrix Rat Gene 1.0 ST Array 10707562 - -
          Affymetrix Rat Gene 1.0 ST Array 10707569 - -
          Affymetrix Rat Gene 1.0 ST Array 10707568 - -
          Affymetrix Rat Gene 1.0 ST Array 10707567 - -
          Affymetrix Rat Gene 1.0 ST Array 10707552 - -
          Affymetrix Rat Gene 1.0 ST Array 10707559 - -
          Affymetrix Rat Gene 1.0 ST Array 10707561 - -
          Affymetrix Rat Gene 1.0 ST Array 10707560 - -
          Affymetrix Rat Gene 1.0 ST Array 10707556 - -
          Affymetrix Rat Gene 1.0 ST Array 10707553 - -
          Affymetrix Rat Gene 1.0 ST Array 10707563 - -
          Affymetrix Rat Gene 1.0 ST Array 10707557 - -
          Affymetrix Rat Gene 1.0 ST Array 10707550 - -
          Affymetrix Rat Gene 1.0 ST Array 10707555 - -
          Affymetrix Rat Gene 1.1 ST Array 10707548 - -
          Affymetrix Rat Clariom S Assay PSR0100013077.rn.1 TC0100001608.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100013078.rn.1 TC0100001608.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10707548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix