Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410034_m1
          See other AQP1 GE Assays ›
          Gene Symbol:
          Aqp1
          Gene Name:
          aquaporin 1
          Gene Aliases:
          CHIP28
          Chromosome Location:
          Chr.4: 85551503 - 85563683 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410034_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aqp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012778.1 NP_036910.1 3 - 4 689 108
          GenBank mRNA BC090068.1 - 3 - 4 671 108
          L07268.1 - 3 - 4 680 108
          X67948.1 - 3 - 4 689 108
          X70257.1 - 3 - 4 629 108
          X71069.1 - 3 - 4 640 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000974.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aqp1

          Gene Ontology Categories:

          Function(s) Process(es)

          glomerular filtration
          renal water transport
          cGMP biosynthetic process
          potassium ion transport
          water transport
          cell volume homeostasis
          hyperosmotic response
          response to xenobiotic stimulus
          response to hormone
          cellular water homeostasis
          positive regulation of lamellipodium assembly
          positive regulation of epithelial cell migration
          carbon dioxide transport
          ammonium transport
          glycerol transport
          sensory perception of pain
          cellular homeostasis
          lateral ventricle development
          water homeostasis
          pancreatic juice secretion
          nitric oxide transport
          positive regulation of cell migration
          establishment or maintenance of actin cytoskeleton polarity
          secretion by cell
          cerebrospinal fluid secretion
          secretory granule organization
          cellular response to UV
          transepithelial water transport
          carbon dioxide transmembrane transport
          wound healing
          odontogenesis
          hyperosmotic salinity response
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to estrogen
          lipid digestion
          positive regulation of angiogenesis
          positive regulation of saliva secretion
          positive regulation of fibroblast proliferation
          camera-type eye morphogenesis
          multicellular organismal water homeostasis
          corticotropin secretion
          renal water absorption
          cellular response to hydrogen peroxide
          cellular response to inorganic substance
          cellular response to mechanical stimulus
          cellular response to copper ion
          cellular response to mercury ion
          cellular response to retinoic acid
          cellular response to cAMP
          cellular response to hypoxia
          cellular response to salt stress
          cellular hyperosmotic response
          cellular response to dexamethasone stimulus
          cellular response to nitric oxide
          potassium ion transmembrane transport
          metanephric descending thin limb development
          metanephric proximal straight tubule development
          metanephric proximal convoluted tubule segment 2 development
          metanephric glomerulus vasculature development
          ammonium transmembrane transport
          cation transmembrane transport
          intracellular cGMP-activated cation channel activity
          potassium channel activity
          water transmembrane transporter activity
          protein binding
          ammonium transmembrane transporter activity
          potassium ion transmembrane transporter activity
          glycerol transmembrane transporter activity
          water channel activity
          glycerol channel activity
          transmembrane transporter activity
          nitric oxide transmembrane transporter activity
          carbon dioxide transmembrane transporter activity
          ephrin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P299247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117284 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299247 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299247 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P299247 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299247 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700373154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100559 - -
          Agilent Rat cDNA Microarray (G4105A) 700373154 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855706 - -
          Affymetrix Rat Gene 1.0 ST Array 10855702 - -
          Affymetrix Rat Gene 1.0 ST Array 10855703 - -
          Affymetrix Rat Gene 1.0 ST Array 10855704 - -
          Affymetrix Rat Gene 1.0 ST Array 10855705 - -
          Affymetrix Rat Gene 1.1 ST Array 10855701 - -
          Rat Genome 230 2.0 Array 1369625_PM_at - -
          Rat Genome 230 2.0 Array 1369625_at - -
          Rat Genome 230 2.0 Array 1387651_PM_at - -
          Rat Genome 230 2.0 Array 1387651_at - -
          Affymetrix HT Rat Focus Array 1369625_at - -
          Affymetrix HT Rat Focus Array 1369625_PM_at - -
          Affymetrix HT Rat Focus Array 1387651_PM_at - -
          Affymetrix HT Rat Focus Array 1387651_at - -
          Affymetrix Rat Clariom S Assay PSR0400006963.rn.1 TC0400000974.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006964.rn.1 TC0400000974.rn.1 RTA TC
          Rat Expression Set 230 1387651_at - -
          Rat Expression Set 230 1369625_at - -
          Rat Expression Set 230 1369625_PM_at - -
          Rat Expression Set 230 1387651_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855701 - -
          Rat Genome U34 Set X67948_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21450146 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358325 - -
          RatRef-12 Expression BeadChip ILMN_53187 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix