Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166342.1 | NP_001159814.1 | 10 - 11 | 2283 | 127 | |
XM_008773406.1 | XP_008771628.1 | 9 - 10 | 1548 | 127 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |