Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01411215_m1
          See other PDZD8 GE Assays ›
          Gene Symbol:
          Pdzd8
          Gene Name:
          PDZ domain containing 8
          Gene Aliases:
          Pdzk8
          Chromosome Location:
          Chr.1: 280458041 - 280515650 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01411215_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdzd8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107446.1 NP_001100916.1 4 - 5 1491 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007970.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          regulation of cell morphogenesis
          intracellular signal transduction
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P182739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150532 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134902 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P182739 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P182739 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P182739 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P182739 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P340915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13010 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731134 - -
          Affymetrix Rat Gene 1.0 ST Array 10731133 - -
          Affymetrix Rat Gene 1.0 ST Array 10731131 - -
          Affymetrix Rat Gene 1.0 ST Array 10731135 - -
          Affymetrix Rat Gene 1.0 ST Array 10731132 - -
          Affymetrix Rat Gene 1.1 ST Array 10731130 - -
          Rat Genome 230 2.0 Array 1390799_PM_at - -
          Rat Genome 230 2.0 Array 1379650_at - -
          Rat Genome 230 2.0 Array 1390799_at - -
          Rat Genome 230 2.0 Array 1379650_PM_at - -
          Affymetrix HT Rat Focus Array 1390799_PM_at - -
          Affymetrix HT Rat Focus Array 1390799_at - -
          Affymetrix Rat Clariom S Assay PSR0100064235.rn.1 TC0100007970.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100064234.rn.1 TC0100007970.rn.1 RTA TC
          Rat Expression Set 230 1390799_at - -
          Rat Expression Set 230 1379650_PM_at - -
          Rat Expression Set 230 1390799_PM_at - -
          Rat Expression Set 230 1379650_at - -
          Affymetrix Rat Gene 1.0 ST Array 10731130 - -
          Rat Genome U34 Set rc_AI177748_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20960620 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370380 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0