Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asmtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105915.2 | NP_001099385.2 | 6 - 7 | 594 | 96 | |
XM_006249001.2 | XP_006249063.1 | 7 - 8 | 711 | 96 | ||
GenBank mRNA | BC160824.1 | - | 5 - 6 | 935 | 96 | |
BP484213.1 | - | 1 - 2 | 135 | 96 | ||
FM053536.1 | - | 4 - 5 | 263 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |