Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413048_m1
          See other PGM2 GE Assays ›
          Gene Symbol:
          Pgm2
          Gene Name:
          phosphoglucomutase 2
          Gene Aliases:
          Pgm1
          Chromosome Location:
          Chr.14: 45829446 - 45863880 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413048_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pgm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106007.1 NP_001099477.1 7 - 8 980 58
          XM_006250996.2 XP_006251058.1 8 - 9 1083 58
          XM_006250997.2 XP_006251059.1 9 - 10 1190 58
          GenBank mRNA BC160893.1 - 8 - 9 1080 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001800.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001106007.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen biosynthetic process
          glucose metabolic process
          biological_process
          galactose catabolic process
          magnesium ion binding
          molecular_function
          phosphoglucomutase activity
          phosphopentomutase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P028446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64862 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96756 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028446 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028446 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P028446 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P455982 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10776939 - -
          Affymetrix Rat Gene 1.0 ST Array 10776948 - -
          Affymetrix Rat Gene 1.0 ST Array 10776941 - -
          Affymetrix Rat Gene 1.0 ST Array 10776950 - -
          Affymetrix Rat Gene 1.0 ST Array 10776949 - -
          Affymetrix Rat Gene 1.0 ST Array 10776943 - -
          Affymetrix Rat Gene 1.0 ST Array 10776940 - -
          Affymetrix Rat Gene 1.0 ST Array 10776947 - -
          Affymetrix Rat Gene 1.0 ST Array 10776944 - -
          Affymetrix Rat Gene 1.0 ST Array 10776952 - -
          Affymetrix Rat Gene 1.0 ST Array 10776946 - -
          Affymetrix Rat Gene 1.0 ST Array 10776951 - -
          Affymetrix Rat Gene 1.0 ST Array 10776942 - -
          Affymetrix Rat Gene 1.0 ST Array 10776945 - -
          Affymetrix Rat Gene 1.1 ST Array 10776938 - -
          Rat Genome 230 2.0 Array 1373769_at - -
          Rat Genome 230 2.0 Array 1373769_PM_at - -
          Affymetrix HT Rat Focus Array 1373769_at - -
          Affymetrix HT Rat Focus Array 1373769_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400013278.rn.1 TC1400001800.rn.1 RTA TC
          Rat Expression Set 230 1373769_at - -
          Rat Expression Set 230 1373769_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10776938 - -
          Rat Genome U34 Set rc_AI176913_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21003956 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355760 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0