Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01413886_m1
          See other CCNG2 GE Assays ›
          Gene Symbol:
          Ccng2
          Gene Name:
          cyclin G2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 16267572 - 16276068 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01413886_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccng2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105725.2 NP_001099195.1 6 - 7 889 78
          GenBank mRNA BC169085.1 - 6 - 7 889 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001458.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          regulation of cell cycle

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P203665 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P715070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139813 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P715070 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P203665 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P203665 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P715070 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P715070 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P203665 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P203665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775766 - -
          Affymetrix Rat Gene 1.0 ST Array 10775763 - -
          Affymetrix Rat Gene 1.0 ST Array 10775767 - -
          Affymetrix Rat Gene 1.0 ST Array 10775764 - -
          Affymetrix Rat Gene 1.0 ST Array 10775765 - -
          Affymetrix Rat Gene 1.0 ST Array 10775762 - -
          Affymetrix Rat Gene 1.0 ST Array 10775768 - -
          Affymetrix Rat Gene 1.1 ST Array 10775761 - -
          Rat Genome 230 2.0 Array 1371953_PM_at - -
          Rat Genome 230 2.0 Array 1371953_at - -
          Affymetrix HT Rat Focus Array 1371953_at - -
          Affymetrix HT Rat Focus Array 1371953_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400010763.rn.1 TC1400001458.rn.1 RTA TC
          Rat Expression Set 230 1371953_at - -
          Rat Expression Set 230 1371953_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775761 - -
          Rat Genome U34 Set rc_AI171262_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21954151 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368551 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0