Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01417812_m1
          See other TIPIN GE Assays ›
          Gene Symbol:
          Tipin Tipinl1
          Gene Name:
          timeless interacting protein
          TIMELESS interacting protein like 1
          Gene Aliases:
          RGD1564084
          -
          Chromosome Location:
          Chr.8: 69753363 - 69768640 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 173775 - 174966 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01417812_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tipin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025287.1 NP_001020458.1 6 - 7 706 76
          GenBank mRNA BC097351.1 - 6 - 7 706 76
          Gene Symbol: Tipinl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002730066.4 XP_002730112.1 1 747 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tipin
          Click here to find siRNAs for Gene Tipinl1

          Gene Ontology Categories:

          Function(s)

          DNA replication checkpoint signaling
          DNA damage checkpoint signaling
          cellular response to DNA damage stimulus
          positive regulation of cell population proliferation
          response to UV
          mitotic intra-S DNA damage checkpoint signaling
          cell cycle phase transition
          replication fork protection

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015577 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P073378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9785 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015577 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073378 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073378 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015577 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015577 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P073378 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P179325 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17640 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10930532 - -
          Affymetrix Rat Gene 1.0 ST Array 10930533 - -
          Affymetrix Rat Gene 1.1 ST Array 10930531 - -
          Rat Genome 230 2.0 Array 1375572_PM_at - -
          Rat Genome 230 2.0 Array 1375572_at - -
          Affymetrix HT Rat Focus Array 1375572_PM_at - -
          Affymetrix HT Rat Focus Array 1375572_at - -
          Affymetrix Rat Clariom S Assay PSR0800006434.rn.1 TC0800000921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800006435.rn.1 TC0800000921.rn.1 RTA TC
          Rat Expression Set 230 1375572_at - -
          Rat Expression Set 230 1375572_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10930531 - -
          Rat Genome U34 Set rc_AI104493_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20717455 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364892 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530503 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0