Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418644_m1
          See other ZBTB16 GE Assays ›
          Gene Symbol:
          Zbtb16
          Gene Name:
          zinc finger and BTB domain containing 16
          Gene Aliases:
          Lx, Plzf, Zfp145, Znf145
          Chromosome Location:
          Chr.8: 52985444 - 53146765 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418644_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013181.1 NP_001013199.1 5 - 6 1785 65
          GenBank mRNA AB568265.1 - 4 - 5 583 65
          AY781102.1 - 5 - 6 1785 65
          AY781103.1 - 5 - 6 1785 65
          AY781104.1 - 5 - 6 1776 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002484.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb16

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          skeletal system development
          mesonephros development
          regulation of transcription, DNA-templated
          central nervous system development
          negative regulation of cell population proliferation
          embryonic pattern specification
          anterior/posterior pattern specification
          hemopoiesis
          embryonic limb morphogenesis
          positive regulation of chondrocyte differentiation
          protein localization to nucleus
          embryonic hindlimb morphogenesis
          forelimb morphogenesis
          hindlimb morphogenesis
          embryonic digit morphogenesis
          positive regulation of apoptotic process
          positive regulation of fat cell differentiation
          negative regulation of myeloid cell differentiation
          positive regulation of ossification
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          male germ-line stem cell asymmetric division
          positive regulation of NK T cell differentiation
          cartilage development
          positive regulation of cartilage development
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          protein binding
          protein C-terminus binding
          protein domain specific binding
          identical protein binding
          protein homodimerization activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P366142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680940 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130374 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77192 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3193 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76716 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103944 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P366142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P366142 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P366142 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P189428 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P366142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10675 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27840 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 206 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93628 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13105 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3066 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917117 - -
          Affymetrix Rat Gene 1.0 ST Array 10917121 - -
          Affymetrix Rat Gene 1.0 ST Array 10917120 - -
          Affymetrix Rat Gene 1.0 ST Array 10917122 - -
          Affymetrix Rat Gene 1.0 ST Array 10917118 - -
          Affymetrix Rat Gene 1.0 ST Array 10917119 - -
          Affymetrix Rat Gene 1.1 ST Array 10917116 - -
          Affymetrix Rat Clariom S Assay PSR0800019969.rn.1 TC0800002484.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019970.rn.1 TC0800002484.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800019968.rn.1 TC0800002484.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10917116 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20991619 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649805 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix