Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423189_g1
          See other TMEM189 GE Assays ›
          Gene Symbol:
          Tmem189
          Gene Name:
          transmembrane protein 189
          Gene Aliases:
          Kua
          Chromosome Location:
          Chr.3: 164376621 - 164388213 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423189_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem189
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113752.1 NP_001107224.1 4 - 5 706 94
          GenBank mRNA BC158660.1 - 4 - 5 706 94

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P270812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152855 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P270812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P270812 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P270812 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P270812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851965 - -
          Affymetrix Rat Gene 1.0 ST Array 10851966 - -
          Affymetrix Rat Gene 1.0 ST Array 10851969 - -
          Affymetrix Rat Gene 1.0 ST Array 10851967 - -
          Affymetrix Rat Gene 1.0 ST Array 10851968 - -
          Affymetrix Rat Gene 1.0 ST Array 10851970 - -
          Affymetrix Rat Gene 1.1 ST Array 10851964 - -
          Rat Genome 230 2.0 Array 1375341_PM_at - -
          Rat Genome 230 2.0 Array 1375341_at - -
          Rat Expression Set 230 1375341_at - -
          Rat Expression Set 230 1375341_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851964 - -
          Rat Genome U34 Set rc_AA943564_at - -
          Rat Genome U34 Set AA942829_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22095681 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530432 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366790 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0