Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01424675_m1
          See other LITAF GE Assays ›
          Gene Symbol:
          Litaf
          Gene Name:
          lipopolysaccharide-induced TNF factor
          Gene Aliases:
          EET-1, Pig7
          Chromosome Location:
          Chr.10: 4753546 - 4763272 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01424675_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Litaf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105735.2 NP_001099205.1 2 - 3 398 62
          GenBank mRNA BC166851.1 - 2 - 3 398 62
          U53184.1 - 435 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000056.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine production
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          signal transduction
          aging
          response to lipopolysaccharide
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          cellular response to lipopolysaccharide
          negative regulation of NIK/NF-kappaB signaling
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P519620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146339 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P519620 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P519620 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P519620 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100975 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P519620 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701586981 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700252828 - -
          Agilent Rat cDNA Microarray (G4105A) 700252828 - -
          Agilent Rat cDNA Microarray (G4105A) 701586981 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731494 - -
          Affymetrix Rat Gene 1.0 ST Array 10731497 - -
          Affymetrix Rat Gene 1.0 ST Array 10731496 - -
          Affymetrix Rat Gene 1.0 ST Array 10731495 - -
          Affymetrix Rat Gene 1.1 ST Array 10731493 - -
          Rat Genome 230 2.0 Array 1370928_PM_at - -
          Rat Genome 230 2.0 Array 1370928_at - -
          Affymetrix HT Rat Focus Array 1370928_at - -
          Affymetrix HT Rat Focus Array 1370928_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000000626.rn.1 TC1000000056.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000000625.rn.1 TC1000000056.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10731493 - -
          Rat Genome U34 Set U53184_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21284927 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357137 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix