Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426423_m1
          See other ZCCHC2 GE Assays ›
          Gene Symbol:
          Zcchc2
          Gene Name:
          zinc finger, CCHC domain containing 2
          Gene Aliases:
          Tnfrsf11a
          Chromosome Location:
          Chr.13: 25966428 - 26014926 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426423_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zcchc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001122677.2 NP_001116149.2 5 - 6 1686 134
          NM_001271042.1 NP_001257971.1 5 - 6 1686 134
          XM_006249625.2 XP_006249687.1 5 - 6 1681 134
          XM_006249626.2 XP_006249688.1 5 - 6 1681 134
          XM_006249627.2 XP_006249689.1 4 - 5 1029 134
          XM_006249628.2 XP_006249690.1 5 - 6 1101 134
          GenBank mRNA BC100090.1 - 5 - 6 652 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000076.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          zinc ion binding
          phosphatidylinositol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059761 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P653818 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470187 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159677 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130866 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151194 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059761 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P653818 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072078 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059761 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P653818 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072078 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072078 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059761 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P653818 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90675 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80660 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763287 - -
          Affymetrix Rat Gene 1.0 ST Array 10763292 - -
          Affymetrix Rat Gene 1.0 ST Array 10763290 - -
          Affymetrix Rat Gene 1.0 ST Array 10763293 - -
          Affymetrix Rat Gene 1.0 ST Array 10763289 - -
          Affymetrix Rat Gene 1.0 ST Array 10763298 - -
          Affymetrix Rat Gene 1.0 ST Array 10763288 - -
          Affymetrix Rat Gene 1.0 ST Array 10763294 - -
          Affymetrix Rat Gene 1.0 ST Array 10763296 - -
          Affymetrix Rat Gene 1.0 ST Array 10763297 - -
          Affymetrix Rat Gene 1.0 ST Array 10763295 - -
          Affymetrix Rat Gene 1.0 ST Array 10763285 - -
          Affymetrix Rat Gene 1.0 ST Array 10763291 - -
          Affymetrix Rat Gene 1.1 ST Array 10763284 - -
          Affymetrix Rat Clariom S Assay PSR1300000327.rn.1 TC1300000076.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000326.rn.1 TC1300000076.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10763284 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21324258 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367097 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0