Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191599.1 | NP_001178528.1 | 6 - 7 | 1075 | 98 | |
XM_006255424.2 | XP_006255486.1 | 6 - 7 | 1111 | 98 |
Set Membership: |
![]() ![]() ![]() |