Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01432660_g1
          See other LOC100909916 GE Assays ›
          Gene Symbol:
          LOC100909916 RGD1564492 Trappc2l
          Gene Name:
          trafficking protein particle complex subunit 2-like protein-like
          similar to hypothetical protein, 2-6
          trafficking protein particle complex 2-like
          Gene Aliases:
          -
          -
          RGD1304906
          Chromosome Location:
          Chr.5: 98683566 - 98733336 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 6979525 - 7024575 on Build Rnor_6.0
          Chromosome Location:
          Chr.19: 55423350 - 55427036 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn01432660_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100909916
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006255170.1 XP_006255232.1 2 343 147
          XM_008763843.1 XP_008762065.1 3 442 147
          Gene Symbol: RGD1564492
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008761693.1 XP_008759915.1 2 331 147
          Gene Symbol: Trappc2l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106193.1 NP_001099663.1 3 - 4 321 147
          GenBank mRNA BC166907.1 - 3 - 4 307 147
          BC166917.1 - 3 - 4 307 147
          BC168734.1 - 3 - 4 307 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          endoplasmic reticulum to Golgi vesicle-mediated transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928527 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015662 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015662 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015662 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P629721 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P419625 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P998800 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10808548 - -
          Affymetrix Rat Gene 1.0 ST Array 10808549 - -
          Affymetrix Rat Gene 1.0 ST Array 10808551 - -
          Affymetrix Rat Gene 1.0 ST Array 10808552 - -
          Affymetrix Rat Gene 1.0 ST Array 10808550 - -
          Affymetrix Rat Gene 1.0 ST Array 10809460 - -
          Affymetrix Rat Gene 1.1 ST Array 10808547 - -
          Affymetrix Rat Gene 1.1 ST Array 10809459 - -
          Rat Genome 230 2.0 Array 1367523_at - -
          Rat Genome 230 2.0 Array 1367523_PM_at - -
          Affymetrix HT Rat Focus Array 1367523_at - -
          Affymetrix HT Rat Focus Array 1367523_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900006329.rn.1 TC1900000783.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006326.rn.1 TC1900000783.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006323.rn.1 TC1900000783.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006331.rn.1 TC1900000783.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006332.rn.1 TC1900000783.rn.1 RTA TC
          Rat Expression Set 230 1367523_PM_at - -
          Rat Expression Set 230 1367523_at - -
          Rat Expression Set 230 1367523_PM_at - -
          Rat Expression Set 230 1367523_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809459 - -
          Affymetrix Rat Gene 1.0 ST Array 10808547 - -
          Rat Genome U34 Set rc_AA924092_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21591874 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039934 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353778 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366963 - -
          RatRef-12 Expression BeadChip ILMN_66266 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0