Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435105_mH
          See other SERPINB6 GE Assays ›
          Gene Symbol:
          Serpinb6
          Gene Name:
          serpin peptidase inhibitor, clade B (ovalbumin), member 6
          Gene Aliases:
          Serpinb6a
          Chromosome Location:
          Chr.17: 32154730 - 32209598 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435105_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199085.2 NP_954516.1 5 - 6 684 70
          XM_006253869.1 XP_006253931.1 5 - 6 682 70
          XM_006253870.2 XP_006253932.1 5 - 6 977 70
          XM_006253871.2 XP_006253933.1 5 - 6 797 70
          XM_008771596.1 XP_008769818.1 5 - 6 895 70
          GenBank mRNA BC060594.1 - 5 - 6 688 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000495.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinb6

          Gene Ontology Categories:

          Function(s) Process(es)

          sensory perception of sound
          negative regulation of endopeptidase activity
          cellular response to osmotic stress
          protease binding
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P505393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26722 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44176 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141616 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P505393 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P505393 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P505393 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P505393 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26378 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794837 - -
          Affymetrix Rat Gene 1.0 ST Array 10794841 - -
          Affymetrix Rat Gene 1.0 ST Array 10794838 - -
          Affymetrix Rat Gene 1.0 ST Array 10794842 - -
          Affymetrix Rat Gene 1.0 ST Array 10794840 - -
          Affymetrix Rat Gene 1.0 ST Array 10794839 - -
          Affymetrix Rat Gene 1.1 ST Array 10794836 - -
          Rat Genome 230 2.0 Array 1375003_at - -
          Rat Genome 230 2.0 Array 1375003_PM_at - -
          Affymetrix HT Rat Focus Array 1375003_at - -
          Affymetrix HT Rat Focus Array 1375003_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700003539.rn.1 TC1700000495.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700003538.rn.1 TC1700000495.rn.1 RTA TC
          Rat Expression Set 230 1375003_PM_at - -
          Rat Expression Set 230 1375003_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794836 - -
          Rat Genome U34 Set rc_AA997608_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21228000 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349114 - -
          RatRef-12 Expression BeadChip ILMN_58990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0