Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435492_m1
          See other ENDOG GE Assays ›
          Gene Symbol:
          Endog
          Gene Name:
          endonuclease G
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 8741833 - 8744414 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435492_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Endog
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034938.1 NP_001030110.1 1 - 2 512 163
          GenBank mRNA AB221075.1 - 1 - 2 512 163

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000201.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Endog

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          apoptotic DNA fragmentation
          aging
          response to mechanical stimulus
          response to estradiol
          cellular response to oxidative stress
          response to tumor necrosis factor
          neuron death in response to oxidative stress
          positive regulation of apoptotic process
          response to antibiotic
          cellular response to calcium ion
          cellular response to glucose stimulus
          cellular response to hypoxia
          positive regulation of hydrogen peroxide-mediated programmed cell death
          positive regulation of apoptotic DNA fragmentation
          nucleic acid binding
          nuclease activity
          endonuclease activity
          deoxyribonuclease activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P213133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170096 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P213133 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P213133 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P213133 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P213133 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835153 - -
          Affymetrix Rat Gene 1.0 ST Array 10835151 - -
          Affymetrix Rat Gene 1.0 ST Array 10835152 - -
          Affymetrix Rat Gene 1.1 ST Array 10844136 - -
          Affymetrix Rat Gene 1.1 ST Array 10835150 - -
          Rat Genome 230 2.0 Array 1373782_a_at - -
          Rat Genome 230 2.0 Array 1389816_at - -
          Rat Genome 230 2.0 Array 1373782_PM_a_at - -
          Rat Genome 230 2.0 Array 1389816_PM_at - -
          Affymetrix HT Rat Focus Array 1389816_at - -
          Affymetrix HT Rat Focus Array 1389816_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300002277.rn.1 TC0300000201.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300002276.rn.1 TC0300000201.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300002279.rn.1 TC0300000201.rn.1 RTA TC
          Rat Expression Set 230 1389816_at - -
          Rat Expression Set 230 1389816_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10835150 - -
          Rat Genome U34 Set rc_AA964570_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22166707 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0