Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108660.1 | NP_001102130.1 | 4 - 5 | 651 | 129 | |
XM_006238073.2 | XP_006238135.1 | 7 - 8 | 604 | 129 | ||
XM_006238074.2 | XP_006238136.1 | 7 - 8 | 601 | 129 | ||
XM_006238076.1 | XP_006238138.1 | 5 - 6 | 430 | 129 | ||
XM_006238077.2 | XP_006238139.1 | 5 - 6 | 443 | 129 | ||
XM_006238079.2 | XP_006238141.1 | 5 - 6 | 399 | 129 | ||
XM_006238081.1 | XP_006238143.1 | 2 - 3 | 1096 | 129 | ||
XM_008763627.1 | XP_008761849.1 | 7 - 8 | 559 | 129 | ||
GenBank mRNA | FQ228666.1 | - | 2 - 3 | 231 | 129 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |