Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106901.2 | NP_001100371.2 | 5 - 6 | 1838 | 101 | |
XM_006244756.2 | XP_006244818.1 | 5 - 6 | 1845 | 101 | ||
XM_006244757.2 | XP_006244819.1 | 5 - 6 | 1848 | 101 |
Set Membership: |
![]() ![]() ![]() ![]() |