Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442167_g1
          See other TOMM7 GE Assays ›
          Gene Symbol:
          Tomm7
          Gene Name:
          translocase of outer mitochondrial membrane 7 homolog (yeast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 7835949 - 7842790 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442167_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tomm7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135174.1 NP_001128646.1 2 - 3 207 103
          GenBank mRNA CB693792.1 - 2 - 3 217 103
          CB698700.1 - 2 - 3 184 103
          FQ221229.1 - 2 - 3 212 103
          FQ223031.1 - 2 - 3 213 103
          FQ223270.1 - 2 - 3 219 103
          FQ224596.1 - 2 - 3 211 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000122.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          protein import into mitochondrial matrix

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P791872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4400 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P791872 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P791872 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P791872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852966 - -
          Affymetrix Rat Gene 1.0 ST Array 10852965 - -
          Affymetrix Rat Gene 1.0 ST Array 10878271 - -
          Affymetrix Rat Gene 1.1 ST Array 10852964 - -
          Affymetrix Rat Gene 1.1 ST Array 10878270 - -
          Rat Genome 230 2.0 Array 1389544_PM_at - -
          Rat Genome 230 2.0 Array 1389544_at - -
          Affymetrix HT Rat Focus Array 1389544_PM_at - -
          Affymetrix HT Rat Focus Array 1389544_at - -
          Affymetrix Rat Clariom S Assay PSR0400000911.rn.1 TC0400000122.rn.1 RTA TC
          Rat Expression Set 230 1389544_at - -
          Rat Expression Set 230 1389544_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852964 - -
          Rat Genome U34 Set rc_AI137062_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix